Orders LinkContactUsLinkComponent
Masterflex® Pressure Booster Pump Systems, Avantor®
Supplier:   VWR International
Masterflex® Pressure Booster Pump Systems, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Pressure Booster Pump Systems, Avantor®
2 Options Available Below
Product Details & Documents
Pump system automatically adjusts speed to maintain set pressure.
  • Maximum flow range of 25 to 100 GPM│NEMA 1 or NEMA 4 variable frequency drive controller with corresponding main disconnect
  • Three-phase input line voltage (single-phase systems are available)
Pump supply booster systems feature a stainless steel centrifugal pump. System is controlled by the variable frequency drive (VFD) which features an LCD and adjustable pressure range; choose the VFD with either a NEMA 1 or NEMA 4 rated enclosure and main disconnect.
System has a small footprint of approximately 30" x 30" (76.2 x 76.2 cm), and has a 1.5" NPT(F) suction and discharge connection. Piping, valve, and drain bodies are made from brass to meet most needs.
Additional options are available: stainless steel piping upgrade, suction check valve (bronze), unions (bronze), low suction shutoff, and low suction shutoff with alarm. If your facility has only single phase voltage, a custom unit can be made to match. Please contact our Application Specialists for details.
Please contact your local Avantor office to customize a system suited to your needs.
 : Includes pump, VFD controller, bladder tank, suction and discharge gauges, discharge check valve, backup relief valve, and necessary piping and drain.
Specifications for Product Family
Specifications
  • Model:
  • 70794-04
  • 70794-22
  • 70794-24
  • 70794-44
  • 70794-64
  • Max. pressure:
  • 4.3 bar (63 psi)
  • 3 bar (44 psi)
  • 3.9 bar (56 psi)
  • 4.3 bar (63 psi)
  • 3.9 bar (56 psi)
  • Motor enclosure type:
  • NEMA 1
  • NEMA 1
  • NEMA 1
  • NEMA 4
  • NEMA 4
  • Motor hp:
  • 2
  • 3
  • 3
  • 2
  • 3
  • Wetted materials:
  • SS pump, brass piping/valves/drain
  • SS pump, brass piping/valves/drain
  • SS pump, brass piping/valves/drain
  • SS pump, brass piping/valves/drain
  • SS pump, brass piping/valves/drain
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Maximum Flow Rate
    Availability
    Price
  • MFLX70794-04
    Pump Booster System, 38 gal./min or 63 PSI, 2 HP, NEMA 1 enclosure
    Pump Booster System, 38 gal./min or 63 PSI, 2 HP, NEMA 1 enclosure
    143846 ml/min (38 gal./min)
    Discontinued
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70794-04
    Model
    70794-04
    Max. pressure
    4.3 bar (63 psi)
    Motor enclosure type
    NEMA 1
    Motor hp
    2
    Wetted materials
    SS pump, brass piping/valves/drain
    Warranty
    1 year
    CE Compliant
  • MFLX70794-22
    Pump Booster System, 93 gal./min or 44 PSI, 3 HP, NEMA 1 enclosure
    Pump Booster System, 93 gal./min or 44 PSI, 3 HP, NEMA 1 enclosure
    352043 ml/min (93 gal./min)
    Discontinued
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70794-22
    Model
    70794-22
    Max. pressure
    3 bar (44 psi)
    Motor enclosure type
    NEMA 1
    Motor hp
    3
    Wetted materials
    SS pump, brass piping/valves/drain
    Warranty
    1 year
    CE Compliant
  • MFLX70794-24
    Pump Booster System, 100 gal./min or 56 PSI, 3 HP, NEMA 1 enclosure
    Pump Booster System, 100 gal./min or 56 PSI, 3 HP, NEMA 1 enclosure
    378541 ml/min (100 gal./min)
    Discontinued
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70794-24
    Model
    70794-24
    Max. pressure
    3.9 bar (56 psi)
    Motor enclosure type
    NEMA 1
    Motor hp
    3
    Wetted materials
    SS pump, brass piping/valves/drain
    Warranty
    1 year
    CE Compliant
  • MFLX70794-44
    Pump Booster System, 38 gal./min or 63 PSI, 2 HP, NEMA 4 enclosure
    Pump Booster System, 38 gal./min or 63 PSI, 2 HP, NEMA 4 enclosure
    143846 ml/min (38 gal./min)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $112,910.00
    $112,910.00
    Each (1UN)
       $112,910.00
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70794-44
    Model
    70794-44
    Max. pressure
    4.3 bar (63 psi)
    Motor enclosure type
    NEMA 4
    Motor hp
    2
    Wetted materials
    SS pump, brass piping/valves/drain
    Warranty
    1 year
    CE Compliant
  • MFLX70794-64
    Pump Booster System, 100 gal./min or 56 PSI, 3 HP, NEMA 4 enclosure
    Pump Booster System, 100 gal./min or 56 PSI, 3 HP, NEMA 4 enclosure
    378541 ml/min (100 gal./min)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $118,370.00
    $118,370.00
    Each (1UN)
       $118,370.00
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70794-64
    Model
    70794-64
    Max. pressure
    3.9 bar (56 psi)
    Motor enclosure type
    NEMA 4
    Motor hp
    3
    Wetted materials
    SS pump, brass piping/valves/drain
    Warranty
    1 year
    CE Compliant