Orders LinkContactUsLinkComponent
Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
Certificates
Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
2 Options Available Below
Product Details & Documents
Place motor and controller where convenient with modular design.
  • Flow rate 0.0006 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, number of pump heads, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Reversible motor—purge tubing before or after pumping and pump fluid in either direction
  • Powerful motor lets you stack two pump heads for increased flow rates and for multichannel applications
These I/P® digital pumps give you rugged performance and precise digital control. These full featured drives are ideal for fluid transfer, dispensing, and metering applications in laboratory, process, and industrial environments including pharmaceutical, food & beverage, dairy, chemical, manufacturing, and water treatment.
Precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Select tubing size and enter desired flow rate—drive locks in required rpm.
Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.
Specifications for Product Family
Specifications
  • Model:
  • 07594-00
  • 07594-10
  • Cloud connected:
  • No
  • No
  • Controller height:
  • 140 mm (5.5")
  • 119 mm (4.75")
  • Controller IP rating:
  • IP 33
  • IP 66
  • Controller length:
  • 263 mm (10.3437")
  • 285 mm (10.1562")
  • Controller width:
  • 211 mm (8.3281")
  • 279 mm (11")
  • Control type:
  • Digital Variable Speed
  • Digital Variable Speed
  • Digital dispensing:
  • Yes
  • Yes
  • Min. RPM:
  • 0,1 rpm
  • 0,1 rpm
  • Motor:
  • 280 W
  • 280 W
  • Motor height:
  • 165 mm (6.5")
  • 165 mm (6.5")
  • Motor hp:
  • 3/8
  • 3/8
  • Motor IP rating:
  • IP 33
  • IP 66
  • Motor length:
  • 362 mm (14.25")
  • 362 mm (14.25")
  • Motor width:
  • 178 mm (7")
  • 178 mm (7")
  • Open-head sensor:
  • No
  • No
  • Pump series:
  • I/P
  • I/P
  • Remote I/O:
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Speed:
  • Variable
  • Variable
  • Speed control:
  • ±0.1%
  • ±0.1%
  • Type of pump heads accepted:
  • I/P Standard, I/P Easy-Load, I/P High-Performance
  • I/P Standard, I/P Easy-Load, I/P High-Performance
  • Washdown capable:
  • No
  • Yes
  • Current:
  • 4.5 A
  • 4.5 A
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
Specifications
Product Family Options
US plug models
Product Information
  • Plug Type
    Maximum Flow Rate
    No. of Pump Heads Accepted
    Housing Material
    Max. RPM
    Power
    Availability
    Price
  • MFLX07594-00
    I/P Digital Modular Drive w/ Remote I/O; 650 rpm
    I/P Digital Modular Drive w/ Remote I/O; 650 rpm
    US
    19000 ml/min
    2
    ABS plastic and aluminium controller; painted steel and aluminium drive
    650 rpm
    90 - 260 VAC; 50/60 Hz; 4.5 A
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $120,900.00
    $120,900.00
    Each (1UN)
       $120,900.00
    Supplier Number: 7594-00
    Specifications:
    2 year warranty
    CE Compliant
    CSA Certified
    More
    Specifications
    Cat. No.
    MFLX07594-00
    Model
    07594-00
    Cloud connected
    No
    Controller height
    140 mm (5.5")
    Controller IP rating
    IP 33
    Controller length
    263 mm (10.3437")
    Controller width
    211 mm (8.3281")
    Control type
    Digital Variable Speed
    Digital dispensing
    Yes
    Min. RPM
    0,1 rpm
    Motor
    280 W
    Motor height
    165 mm (6.5")
    Motor hp
    3/8
    Motor IP rating
    IP 33
    Motor length
    362 mm (14.25")
    Motor width
    178 mm (7")
    Open-head sensor
    No
    Pump series
    I/P
    Remote I/O
    0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    ±0.1%
    Type of pump heads accepted
    I/P Standard, I/P Easy-Load, I/P High-Performance
    Washdown capable
    No
    Current
    4.5 A
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
    CSA Certified
  • MFLX07594-10
    I/P Digital Modular Washdown Drive w/ Remote I/O; 650 rpm
    I/P Digital Modular Washdown Drive w/ Remote I/O; 650 rpm
    US
    19000 ml/min
    2
    Painted aluminium controller; painted steel and aluminium drive
    650 rpm
    90 - 260 VAC; 50/60 Hz; 4.5 A
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $153,080.00
    $153,080.00
    Each (1UN)
       $153,080.00
    Supplier Number: 7594-10
    Specifications:
    2 year warranty
    CE Compliant
    IP66 washdown
    More
    Specifications
    Cat. No.
    MFLX07594-10
    Model
    07594-10
    Cloud connected
    No
    Controller height
    119 mm (4.75")
    Controller IP rating
    IP 66
    Controller length
    285 mm (10.1562")
    Controller width
    279 mm (11")
    Control type
    Digital Variable Speed
    Digital dispensing
    Yes
    Min. RPM
    0,1 rpm
    Motor
    280 W
    Motor height
    165 mm (6.5")
    Motor hp
    3/8
    Motor IP rating
    IP 66
    Motor length
    362 mm (14.25")
    Motor width
    178 mm (7")
    Open-head sensor
    No
    Pump series
    I/P
    Remote I/O
    0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    ±0.1%
    Type of pump heads accepted
    I/P Standard, I/P Easy-Load, I/P High-Performance
    Washdown capable
    Yes
    Current
    4.5 A
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
    IP66 washdown
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Extension cable for L/S and I/P digital modular drives
    Extension cable for L/S and I/P digital modular drives
    MFLX77301-82
    9 ft.
    Log in for availability
    Each (9Pie) - $13,540.00
Product Information
  • Catalog #
    Availability
    Price
  • Cable assembly for remote control, DB25(M) connector
    Cable assembly for remote control, DB25(M) connector
    MFLX07523-95
    Log in for availability
    Each (25Pie) - $5,600.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Cable assembly for remote control, 31-pin round connector and 7.9 m (25') cable with stripped wire ends
    Cable assembly for remote control, 31-pin round connector and 7.9 m (25') cable with stripped wire ends
    MFLX07575-80
    7.3 m (24 ft.)
    Log in for availability
    Each (1Pie) - $5,230.00