ContactUsLinkComponent
Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
undefined
Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
  • Modelo:
  • 74000-53
  • 74000-59
  • 74000-49
  • 74000-51
  • Precisión:
  • ±0.1%
  • ±0.1%
  • ±0.1%
  • ±0.1%
  • CSA regulatory approval:
  • Yes
  • Yes
  • Yes
  • Yes
  • Intrinsically safe regulatory approval:
  • IP54
  • IP54
  • IP54
  • IP54
  • Inlet connection:
  • 3⁄4" Tri-Clamp
  • 3⁄4" Tri-Clamp
  • 1⁄4" Tri-Clamp®
  • 1⁄4" Tri-Clamp®
  • Presión máx.:
  • 4 bar (58 psi)
  • 4 bar (58 psi)
  • 4 bar (58 psi)
  • 4 bar (58 psi)
  • Min. temperature:
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • Max. temperature:
  • 80 °C (176 °F)
  • 60 °C (140 °F)
  • 80 °C (176 °F)
  • 60 °C (140 °F)
  • Velocidad mín. de flujo:
  • 0.04 L/min
  • 0.33 L/min
  • 0.02 L/min
  • 0.02 L/min
  • Motor hp:
  • 0.31
  • 0.31
  • 0.09
  • 0.09
  • Min. motor RPM:
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • Outlet connection:
  • 3⁄4" Tri-Clamp
  • 3⁄4" Tri-Clamp
  • 1⁄4" Tri-Clamp®
  • 1⁄4" Tri-Clamp®
  • Salida:
  • Ethernet/IP
  • Ethernet/IP
  • Ethernet/IP
  • Ethernet/IP
  • Wetted materials:
  • 316L SS (housing, valve plate),TPE (diaphragm), EPDM(valves, O-rings)
  • PP (housing, valve plate),TPE (diaphragm), EPDM(valves, O-rings)
  • 316L SS (housing, valve plate),TPE (diaphragm), EPDM(valves, O-rings)
  • PP (housing, valve plate),TPE (diaphragm), EPDM(valves, O-rings)
  • Garantía:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Frequently Bought Together