Masterflex® I/P® Digital Modular Drive Systems, Avantor®
Acerca de este artículo
Place motor and controller where convenient with modular design.
- Flow rate 0.06 to 8 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
- Digital interface with LED display shows pump rpm— confirm speed setting at a glance
- Remote control available using analog communication—voltage, current and contact closure
- Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
- Membrane keypad allows for easy up-front control of speed, direction, and start/stop
I/P® modular drives feature reversible pumping while allowing you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering and critical transfer applications.
2 Options Available Below
Product Details & Documents
Place motor and controller where convenient with modular design.
- Flow rate 0.06 to 8 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
- Digital interface with LED display shows pump rpm— confirm speed setting at a glance
- Remote control available using analog communication—voltage, current and contact closure
- Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
- Membrane keypad allows for easy up-front control of speed, direction, and start/stop
I/P® modular drives feature reversible pumping while allowing you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering and critical transfer applications.
Documents
Specifications for Product Family
Especificaciones
- Catalog No:
- MFLX77962-20
- MFLX77962-30
- Model:
- 77962-20
- 77962-30
- No. of channels:
- 1
- 1
- No. of rollers:
- 3
- 3
- Control type:
- Variable-Speed Digital
- Variable-Speed Digital
- Min. flow rate per channel:
- 60 ml/min
- 60 ml/min
- Max. flow rate per channel:
- 8000 ml/min
- 8000 ml/min
- Min. flow rate:
- 60 ml/min
- 60 ml/min
- Drive IP rating:
- IP 33
- IP 66
- Accuracy:
- ±0.25% (1 rpm at 650 rpm)
- ±0.25% (1 rpm at 650 rpm)
- Anti-drip:
- No
- No
- Cloud connected:
- No
- No
- Digital dispensing:
- No
- No
- Drive:
- 07591-20 (I/P® Digital Modular Drive with Remote Input/Output)
- 07591-30 (I/P® Digital Washdown Modular Drive with Wall-Mount Controller and Remote I/O)
- Duty cycle:
- Continuous
- Continuous
- Min. RPM:
- 6 rpm
- 6 rpm
- Open-head sensor:
- No
- No
- Pump series:
- I/P
- I/P
- Remote I/O:
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- Reversible motor:
- Yes
- Yes
- Tubing:
- 06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
- 06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
- Tubing sizes accepted:
- I/P® 26, 73, 82
- I/P® 26, 73, 82
- Washdown capable:
- No
- Yes
- Current:
- 4.4 A @ 115 V; 2.2 A @ 230 V
- 4.4 A @ 115 V; 2.2 A @ 230 V
- Frequency:
- 50/60 Hz
- 50/60 Hz
- Voltage:
- 90 - 260 VAC
- 90 - 260 VAC
- Warranty:
- 2 years
- 2 years
Especificaciones
Product Family Options
US plug models
Información del producto
- Plug TypeMaximum Flow RateHeightLengthWidthPump HeadPowerMax. RPMHousing MaterialAvailabilityPrice
- MFLX77962-20I/P Precision Modular Pump w/ Easy-Load Head; 650 rpmI/P Precision Modular Pump w/ Easy-Load Head; 650 rpmUS8000 ml/min155.7 mm (6.13")353 mm (13.88")139.7 mm (5.5")77602-10 (I/P® Easy-Load® pump head)90 - 260 VAC; 50/60 Hz; 4.4 A @ 115 V; 2.2 A @ 230 V650 rpmABS plastic and aluminium controller; painted steel driveLog in Para disponibilidadEach (1UN)$112,130.00Supplier Number: 77962-20Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77962-20Model77962-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalMin. flow rate per channel60 ml/minMax. flow rate per channel8000 ml/minMin. flow rate60 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 650 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07591-20 (I/P® Digital Modular Drive with Remote Input/Output)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPump seriesI/PRemote I/O4 to 20 mA, 0 to 10 V, start/stop and direction via contact closureReversible motorYesTubing06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)Tubing sizes acceptedI/P® 26, 73, 82Washdown capableNoCurrent4.4 A @ 115 V; 2.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years2 year warranty - MFLX77962-30I/P Precision Modular Washdown Pump w/ Easy-Load Head; 650 rpmI/P Precision Modular Washdown Pump w/ Easy-Load Head; 650 rpmUS8000 ml/min165.1 mm (6.5")362 mm (14.25")177.8 mm (7")77602-10 (I/P® Easy-Load® pump head)90 - 260 VAC; 50/60 Hz; 4.4 A @ 115 V; 2.2 A @ 230 V650 rpmPainted aluminium controller; painted steel and aluminium driveLog in Para disponibilidadEach (1UN)$140,600.00Supplier Number: 77962-30Especificaciones:2 year warrantyCE CompliantIP66 washdown
More
Specifications
Cat. No.MFLX77962-30Model77962-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalMin. flow rate per channel60 ml/minMax. flow rate per channel8000 ml/minMin. flow rate60 ml/minDrive IP ratingIP 66Accuracy±0.25% (1 rpm at 650 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07591-30 (I/P® Digital Washdown Modular Drive with Wall-Mount Controller and Remote I/O)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPump seriesI/PRemote I/O4 to 20 mA, 0 to 10 V, start/stop and direction via contact closureReversible motorYesTubing06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)Tubing sizes acceptedI/P® 26, 73, 82Washdown capableYesCurrent4.4 A @ 115 V; 2.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years2 year warrantyCE CompliantIP66 washdown