Orders LinkContactUsLinkComponent
Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
Certificates
Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
3 Options Available Below
Product Details & Documents
Make the most of your Masterflex® Open-Head Sensor with these accessories and replacement parts.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
Specifications for Product Family
Especificaciones
  • Warranty:
  • 2 years
  • 1 year
  • 2 years
Especificaciones
Product Family Options
Información del producto
  • Availability
    Price
  • MFLX07528-82
    Replacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
    Replacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $851.00
    $851.00
    Each (1UN)
       $851.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07528-82
    Warranty
    2 years
  • MFLX77097-02
    Power Cable for Masterflex Open-Head Sensor Interface; 115 V. 6 ft (1.8 m) L
    Power Cable for Masterflex Open-Head Sensor Interface; 115 V. 6 ft (1.8 m) L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $812.00
    $812.00
    Each (1UN)
       $812.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX77097-02
    Warranty
    1 year
  • MFLX77097-20
    Extension Cable for Masterflex Open-Head Sensor Interface; 6 ft (1,8 m) L
    Extension Cable for Masterflex Open-Head Sensor Interface; 6 ft (1,8 m) L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,770.00
    $2,770.00
    Each (1UN)
       $2,770.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX77097-20
    Warranty
    2 years