Masterflex® Open-Head Sensor Retrofit System, Avantor®
Open-Head Sensor Retrofit System Controller, 2
Catalog # MFLX77097-00
CE Compliant
ETL listed
ISO9001
Supplier: Avantor Fluid Handling
Supplier #: 77097-00
Masterflex® Open-Head Sensor Retrofit System, Avantor®
Catalog # MFLX77097-00
Supplier: Avantor Fluid Handling
CE Compliant
ETL listed
ISO9001
Jump to:
Product Details & Documents
Instantly shuts down the drive if the pump head is opened during operation.
- Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
- Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
- Controller accepts input signals from up to two pump heads
- Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
- Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
Specifications
- Description:Open-Head Sensor Retrofit System Controller
- No. of Pump Heads Accepted:2
- Cat. No.:MFLX77097-00
- Warranty:2 years
- Drive requirements:Compatible with all I/P drives and L/S drives that accept the Easy-Load II pump head
- Type of pump heads accepted:Use only with I/P Easy-Load pump head 77602-30 and L/S Easy-Load II pump heads 77200-30 and 77200-32
- Controller IP rating:IP 66
Accessories
Product Information
- Catalog #AvailabilityPrice
- Power Cable for Masterflex Open-Head Sensor Interface; 115 V. 6 ft (1.8 m) LPower Cable for Masterflex Open-Head Sensor Interface; 115 V. 6 ft (1.8 m) LMFLX77097-02Log in for availabilityEach (1UN) - $812.00
Product Information
- Catalog #AvailabilityPrice
- Extension Cable for Masterflex Open-Head Sensor Interface; 6 ft (1,8 m) LExtension Cable for Masterflex Open-Head Sensor Interface; 6 ft (1,8 m) LMFLX77097-20Log in for availabilityEach (1UN) - $2,770.00
Product Information
- Catalog #AvailabilityPrice
- Replacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped DrivesReplacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped DrivesMFLX07528-82Log in for availabilityEach (1UN) - $851.00