Masterflex® Batch Control Drum Pump Systems, Avantor®
Acerca de este artículo
Save money by accurately dispensing expensive fluids — to ±0.5%.
- Flow rate 25 GMP for all models
- Automatically dispense preset volumes of fluid from drums, tanks, and storage containers
- Control transfer of highly corrosive fluids directly from the storage container
- Save up to seven preset volumes in the automatic batch memory
Batch control drum pumps are ideal for applications in chemical packaging, waste water treatment, chemical manufacturing, plating, semi-conductor, pharmaceutical and agricultural industries.
These pumps are engineered to control, measure, and dispense controlled volumes of fluid from drums, tanks and small storage vessels. Simply select the desired volume to transfer, press the enter key and watch the pump accurately dispense the fluid.
Pumps are available with either polypropylene or PVDF pump tubes for chemical compatibility with your fluid. Flow rate can be displayed in gallons, Imperial gallons, liters, milliliters, or cubic meters per minute, second, or hour. Volume can be displayed in gallons, Imperial gallons, liters, milliliters, or cubic meters. Select from five different languages for display: English, German, French, Italian, or Spanish. Flowmeters also feature a re-settable totalizer, EEprom electronics, and a paddle wheel design for continuous flow measurement of neutral and aggressive solid-free fluids.
Delivery information: Includes motor, pump tube, batch control meter, check valve, and hose barb.
4 Options Available Below
Product Details & Documents
Save money by accurately dispensing expensive fluids — to ±0.5%.
- Flow rate 25 GMP for all models
- Automatically dispense preset volumes of fluid from drums, tanks, and storage containers
- Control transfer of highly corrosive fluids directly from the storage container
- Save up to seven preset volumes in the automatic batch memory
Batch control drum pumps are ideal for applications in chemical packaging, waste water treatment, chemical manufacturing, plating, semi-conductor, pharmaceutical and agricultural industries.
These pumps are engineered to control, measure, and dispense controlled volumes of fluid from drums, tanks and small storage vessels. Simply select the desired volume to transfer, press the enter key and watch the pump accurately dispense the fluid.
Pumps are available with either polypropylene or PVDF pump tubes for chemical compatibility with your fluid. Flow rate can be displayed in gallons, Imperial gallons, liters, milliliters, or cubic meters per minute, second, or hour. Volume can be displayed in gallons, Imperial gallons, liters, milliliters, or cubic meters. Select from five different languages for display: English, German, French, Italian, or Spanish. Flowmeters also feature a re-settable totalizer, EEprom electronics, and a paddle wheel design for continuous flow measurement of neutral and aggressive solid-free fluids.
Delivery information: Includes motor, pump tube, batch control meter, check valve, and hose barb.
Specifications for Product Family
Especificaciones
- Max. temperature:
- 130 °C
- 175 °C
- 130 °C
- 175 °C
- Max. viscosity:
- 300 cP
- 300 cP
- 300 cP
- 300 cP
- Motor type:
- ODP
- TEFC
- ODP
- TEFC
- Motor hp:
- 1
- 1
- 1
- 1
- Outlet connection:
- 1" hose barb or 1" NPT thread
- 1" hose barb or 1" NPT thread
- 1" hose barb or 1" NPT thread
- 1" hose barb or 1" NPT thread
- Tube length, inlet:
- 39"
- 39"
- 47"
- 47"
- Wetted materials:
- Polypropylene
- PVDF
- Polypropylene
- PVDF
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
Especificaciones
Product Family Options
Información del producto
- Maximum Flow RatePowerMax. Motor RPMAvailabilityPrice
- MFLX70612-00Batch control drum pump system, PP, 39", 110 VAC, 60 HzBatch control drum pump system, PP, 39", 110 VAC, 60 Hz94.64 L/min (25 gal./min)110 VAC; 60 Hz10000 rpmLog in Para disponibilidadEach (1UN)$115,250.00Especificaciones:
More
Specifications
Cat. No.MFLX70612-00Max. temperature130 °CMax. viscosity300 cPMotor typeODPMotor hp1Outlet connection1" hose barb or 1" NPT threadTube length, inlet39"Wetted materialsPolypropyleneWarranty1 year - MFLX70612-20Batch control drum pump system, PVDF, 39", 110 VAC, 60 HzBatch control drum pump system, PVDF, 39", 110 VAC, 60 Hz94.64 L/min (25 gal./min)110 VAC; 60 Hz10000 rpmLog in Para disponibilidadEach (1UN)$131,630.00Especificaciones:
More
Specifications
Cat. No.MFLX70612-20Max. temperature175 °CMax. viscosity300 cPMotor typeTEFCMotor hp1Outlet connection1" hose barb or 1" NPT threadTube length, inlet39"Wetted materialsPVDFWarranty1 year - MFLX70612-10Batch control drum pump system, PP, 47", 110 VAC, 60 HzBatch control drum pump system, PP, 47", 110 VAC, 60 Hz94.64 L/min (25 gal./min)110 VAC; 60 Hz10000 rpmLog in Para disponibilidadEach (1UN)$97,890.00Especificaciones:
More
Specifications
Cat. No.MFLX70612-10Max. temperature130 °CMax. viscosity300 cPMotor typeODPMotor hp1Outlet connection1" hose barb or 1" NPT threadTube length, inlet47"Wetted materialsPolypropyleneWarranty1 year - MFLX70612-30Batch control drum pump system, PVDF, 47", 110 VAC, 60 HzBatch control drum pump system, PVDF, 47", 110 VAC, 60 Hz94.64 L/min (25 gal./min)110 VAC; 60 Hz10000 rpmLog in Para disponibilidadEach (1UN)$132,210.00Especificaciones:
More
Specifications
Cat. No.MFLX70612-30Max. temperature175 °CMax. viscosity300 cPMotor typeTEFCMotor hp1Outlet connection1" hose barb or 1" NPT threadTube length, inlet47"Wetted materialsPVDFWarranty1 year
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Replacement Wall Mounting Bracket for Drum PumpsReplacement Wall Mounting Bracket for Drum PumpsMFLX70604-90Log in Para disponibilidadEach (1UN) - $1,390.00
Información del producto
- Catalog #AvailabilityPrice
- Replacement Barrel Adapter for Drum Pumps, PPReplacement Barrel Adapter for Drum Pumps, PPMFLX70604-92Log in Para disponibilidadEach (1UN) - $1,370.00
Información del producto
- Catalog #MaterialAvailabilityPrice
- Strainer for Drum Pumps, PP
Información del producto
- Catalog #AvailabilityPrice
- Dispensing Nozzle, PVDF, with 6-ft of PVC hoseDispensing Nozzle, PVDF, with 6-ft of PVC hoseMFLX70604-98Log in Para disponibilidadEach (1UN) - $20,480.00