Masterflex® Transfer Tubing, PVC, Avantor®
Stop oxidation and bacteria problems.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
This tubing is extremely flexible and transparent and is resistant to oxidation, bacteria and a broad range of chemicals. PVC tubing meets FDA CFR Title 21 177.101.
- Environmentally Preferable:
- Catalog No:
- MFLX96605-15
- MFLX96605-25
- MFLX96605-10
- Modelo:
- 96605-15
- 96605-25
- 96605-10
- Clarity:
- Translucent
- Translucent
- Translucent
- Length per pack:
- 15.2 m (50 ft.)
- 15.2 m (50 ft.)
- 15.2 m (50 ft.)
- Presión máx.:
- 2.75 bar (40 psi)
- 2.75 bar (45 psi)
- 4.5 bar (65 psi)
- Max. temperature:
- 74 °C (165 °F)
- 74 °C (165 °F)
- 74 °C (165 °F)
- Min. temperature:
- –41 °C (–41 °F)
- –41 °C (–41 °F)
- –41 °C (–41 °F)
- Regulatory approvals:
- FDA
- FDA
- FDA
- Sterilisation method autoclave:
- No
- No
- No
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes
- Yes
- Yes
- Working pressure:
- 40 psi
- 45 psi
- 65 psi