Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® LMT-55, Avantor®
Supplier: VWR International
Certificates
About this item
Designed for optimal operation in your Ismatec pumps.
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
The overall length of LMT-55 97618-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX97618-34
- MFLX97618-32
- MFLX97618-31
- MFLX97618-40
- MFLX97618-39
- MFLX97618-38
- MFLX97618-36
- MFLX97618-09
- MFLX97618-46
- MFLX97618-44
- MFLX97618-42
- MFLX97618-10
- MFLX97618-16
- MFLX97618-14
- MFLX97618-49
- MFLX97618-48
- MFLX97618-26
- MFLX97618-24
- MFLX97618-22
- MFLX97618-18
- MFLX97618-30
- MFLX97618-29
- MFLX97618-28
- MFLX97618-27
- Modelo:
- 97618-34
- 97618-32
- 97618-31
- 97618-40
- 97618-39
- 97618-38
- 97618-36
- 97618-09
- 97618-46
- 97618-44
- 97618-42
- 97618-10
- 97618-16
- 97618-14
- 97618-49
- 97618-48
- 97618-26
- 97618-24
- 97618-22
- 97618-18
- 97618-30
- 97618-29
- 97618-28
- 97618-27
- Pump series:
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Regulatory approvals:
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Non-DEHP
- Series:
- Similar to SC0016T
- Similar to SC0015T
- Similar to SC0014T
- Similar to SC0020T
- Similar to SC0019T
- Similar to SC0018T
- Similar to SC0017T
- Similar to SC0188T
- Similar to SC0023T
- Similar to SC0022T
- Similar to SC0021T
- Similar to SC0001T
- Similar to SC0004T
- Similar to SC0003T
- Similar to SC0222T
- Similar to SC0024T
- Similar to SC0009T
- Similar to SC0008T
- Similar to SC0007T
- Similar to SC0005T
- Similar to SC0013T
- Similar to SC0012T
- Similar to SC0011T
- Similar to SC0010T
- Tubes per pack:
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12