Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Versilon™ 2001, Avantor®
Designed for optimal operation in your Ismatec pumps.
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
The overall length of Tygon Chemical 96465-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 381 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
- Environmentally Preferable:
- Modelo:
- 96465-14
- 96465-42
- 96465-36
- 96465-28
- 96465-22
- 96465-48
- Pump series:
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Ismatec
- Regulatory approvals:
- ADCF;FDA;Non-DEHP;RoHS
- ADCF;FDA;Non-DEHP;RoHS
- ADCF;FDA;Non-DEHP;RoHS
- ADCF;FDA;Non-DEHP;RoHS
- ADCF;FDA;Non-DEHP;RoHS
- ADCF;FDA;Non-DEHP;RoHS
- Series:
- Similar to SC0814
- Similar to SC0822
- Similar to SC0820
- Similar to SC0818
- Similar to SC0816
- Similar to SC0824
- Tubes per pack:
- 6
- 6
- 6
- 6
- 6
- 6
Frequently Bought Together