Orders LinkContactUsLinkComponent
Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier:  VWR International
Certificates
Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
3 Options Available Below
Product Details & Documents
Long pumping life with superior flexibility reduces downtime and tubing failure.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Extremely low spallation for reduced fluid contamination
Masterflex® Tygon® E-FLF High-Performance Precision pump tubing is a non-DEHP formulation specially formulated for even longer pumping life with superior flexibility. This tubing features low particulate spallation and no phthalates or BPA, protecting fluid purity. Ideal for food and beverage, cosmetics, and pharmaceutical applications, this tubing complies with FDA, USP Class VI, and EU Foods standards.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Specifications for Product Family
Especificaciones
  • Model:
  • 06440-70
  • 06440-88
  • 06440-89
  • Length per pack:
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • Max. pressure:
  • 2.7 bar (40 psi)
  • 2.4 bar (35 psi)
  • 1.4 bar (20 psi)
  • Min. temperature:
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • Max. temperature:
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • Min. temperature (dynamic):
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • Max. temperature (dynamic):
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Max. vacuum:
  • 660 mm Hg (26" Hg)
  • 660 mm Hg (26" Hg)
  • 610 mm Hg (24" Hg)
  • Pump series:
  • I/P
  • I/P
  • I/P
  • Regulatory approvals:
  • ADCF;EU Food 10/2011;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • ADCF;EU Food 10/2011;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • ADCF;EU Food 10/2011;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • Sterilisation:
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • No
  • No
  • No
  • Suction lift H₂O:
  • 8.8 m (29 ft.)
  • 8.8 m (29 ft.)
  • 8.2 m (27 ft.)
  • Warranty:
  • Out of box
  • Out of box
Especificaciones
Product Family Options
Información del producto
  • Tubing Material
    Tubing int.Ø
    Hose Barb Size
    I/P Masterflex Tubing Size
    Maximum Flow Rate
    Availability
    Price
  • MFLX06440-70
    Tygon E-LFL Pump Tubing, I/P 70, 25 ft.
    Tygon E-LFL Pump Tubing, I/P 70, 25 ft.
    Tygon® S3™ E-LFL
    9.50 mm (0.374")
    0.375"
    70
    8000 ml/min
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25Pie)
    Each (25Pie) - $12,720.00
    $12,720.00
    Each (25Pie)
       $12,720.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06440-70
    Model
    06440-70
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    2.7 bar (40 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    I/P
    Regulatory approvals
    ADCF;EU Food 10/2011;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8.8 m (29 ft.)
    Warranty
    Out of box
    CE certificate
  • MFLX06440-88
    Tygon E-LFL Pump Tubing, I/P 88, 25 ft.
    Tygon E-LFL Pump Tubing, I/P 88, 25 ft.
    Tygon® S3™ E-LFL
    12.70 mm (0.500")
    0.5"
    88
    17000 ml/min
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25Pie)
    Each (25Pie) - $15,700.00
    $15,700.00
    Each (25Pie)
       $15,700.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06440-88
    Model
    06440-88
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    2.4 bar (35 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    I/P
    Regulatory approvals
    ADCF;EU Food 10/2011;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8.8 m (29 ft.)
    Warranty
    CE certificate
  • MFLX06440-89
    Tygon E-LFL Pump Tubing, I/P 89, 25 ft.
    Tygon E-LFL Pump Tubing, I/P 89, 25 ft.
    Tygon® S3™ E-LFL
    15.88 mm (0.625")
    0.625"
    89
    19000 ml/min
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25Pie)
    Each (25Pie) - $16,440.00
    $16,440.00
    Each (25Pie)
       $16,440.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06440-89
    Model
    06440-89
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    1.4 bar (20 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    610 mm Hg (24" Hg)
    Pump series
    I/P
    Regulatory approvals
    ADCF;EU Food 10/2011;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8.2 m (27 ft.)
    Warranty
    Out of box
    CE certificate