Masterflex® Extension Cables for Modular Drives, Avantor®
Acerca de este artículo
Increase modular drive flexibility with extended cable lengths.
- Easily connect to your Masterflex® pump with connectors on both ends
Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2.7 m) to increase installation flexibility. Do not extend more than 25ft.
2 Options Available Below
Product Details & Documents
Increase modular drive flexibility with extended cable lengths.
- Easily connect to your Masterflex® pump with connectors on both ends
Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2.7 m) to increase installation flexibility. Do not extend more than 25ft.
Specifications for Product Family
Especificaciones
Especificaciones
Product Family Options
Información del producto
- LengthAvailabilityPrice
- MFLX07557-51Extension cable for L/S and I/P precision modular drivesExtension cable for L/S and I/P precision modular drives9 ft.Log in Para disponibilidadEach (9Pie)$11,510.00Supplier Number: 7557-51Especificaciones:
More
Specifications
Cat. No.MFLX07557-51Model07557-51Warranty2 years - MFLX77301-82Extension cable for L/S and I/P digital modular drivesExtension cable for L/S and I/P digital modular drives9 ft.Log in Para disponibilidadEach (9Pie)$13,540.00Supplier Number: 77301-82Especificaciones:
More
Specifications
Cat. No.MFLX77301-82Model77301-82Warranty2 years