Masterflex® Remote Control Cable Connectors, Avantor®
Supplier: Avantor Fluid Handling
About this item
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
3 Options Available Below
Product Details & Documents
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
Documents
Masterflex LS MasterSense Drives Pump
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- MFLX07523-94
- MFLX07595-45
- MFLX07595-52
- Model:
- 07523-94
- 07595-45
- 07595-52
- Control type:
- Analog Variable Speed
- Warranty:
- 2 years
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- MFLX07523-94DB25 Male connector; for creating your own cableSupplier Number: 7523-94Specifications:
More
Specifications
Cat. No.MFLX07523-94Model07523-94Control typeAnalog Variable SpeedWarranty2 years - MFLX07595-45DB9 male 9-pin connector for use with drivesSupplier Number: 7595-45Specifications:
More
Specifications
Cat. No.MFLX07595-45Model07595-45Control typeWarranty - MFLX07595-52DB15 male connector for creating cableSupplier Number: 7595-52Specifications:
More
Specifications
Cat. No.MFLX07595-52Model07595-52Control typeWarranty