Masterflex® Panel Mount Lock Nut, Black Nylon, Avantor®
Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 25/PK, Threaded Nut, Straight, Nylon
Catalog # MFLX45509-04
Avantor exclusive
Supplier: VWR International
Masterflex® Panel Mount Lock Nut, Black Nylon, Avantor®
Catalog # MFLX45509-04
Supplier: VWR International
Avantor exclusive
Jump to:
Product Details & Documents
Best for use with panel mount fittings.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Optimize your application with these luer fitting accessories.
Especificaciones
- Connection Type:Threaded Nut
- Flow pattern:Straight
- Description:Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 25/PK
- Fitting Material:Nylon
- Cat. No.:MFLX45509-04
- Warranty:3 months