Masterflex® Sterile Forceps, stainless steel, 4.5" long; 25/pk
Catalog # MFLX75955-13
Supplier: VWR International
Specifications
- Descripción:Sterile Forceps, stainless steel, 4,5" long; 25/pk
- Matriz:Stainless steel
- Referencia:MFLX75955-13
- Longitud total:114 mm (4.5")
- Sterile:+
Specifications
About this item
No more cross-contamination.
- Protect yourself and your samples with disposable forceps
- Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.
- Descripción:Sterile Forceps, stainless steel, 4,5" long; 25/pk
- Matriz:Stainless steel
- Environmentally Preferable:
- Referencia:MFLX75955-13
- Longitud total:114 mm (4.5")
- Sterile:+