Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Supplier: VWR International
Certificates
About this item
Protect equipment with automatic flow rate monitoring via high and low alarms.
- Protect your equipment from damage caused by extreme flow rates
- Features dual-mounted LED receiver/sensors with optical sensors
These 150-mm valved flowmeters feature two noninvasive optical sensors that easily glide up and down both sides of the meter to set high and low flow rates. The meter is ideal for use with an alarm, valve, or other devices to control flow with use of NPN and PNP transistor outputs. The compact design and ease of operation make this meter perfect for laboratory and industrial uses.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX33402-44
- MFLX33402-48
- MFLX33402-14
- MFLX33402-34
- MFLX33402-38
- MFLX33402-40
- MFLX33402-04
- MFLX33402-10
- MFLX33402-12
- Modelo:
- 33402-44
- 33402-48
- 33402-14
- 33402-34
- 33402-38
- 33402-40
- 33402-04
- 33402-10
- 33402-12
- Float material:
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Glass
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Glass
- Max. air flow rate:
- 16737 ml/min
- 45227 ml/min
- 4662 ml/min
- 814 ml/min
- 2313 ml/min
- 4662 ml/min
- 264 ml/min
- 1682 ml/min
- 2313 ml/min
- Repetibilidad:
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year