Masterflex® Vortex Flowmeters with Temperature for Liquids, Avantor®
Acerca de este artículo
Built-in 1000 Ω RTD sensor provides direct temperature measurement of media.
- Deigned with rugged PPA molded construction for enhanced durability
- Excellent media compatibility with ETFE, EPDM, and PPD wetted parts
- Use with low viscosity liquids for laboratory and industrial applications
These economical vortex flowmeters are suitable for water and water glycol-based heat exchange systems. The transmitters work on the vortex shedding flow principle. As flow velocity increases, alternating vortices are formed on each side of the bluff body within the flowtube and travel downstream. The flowmeters utilize piezoelectric sensors embedded in a vane downstream from the bluff to detect the vortices and transfer the sensed vortices into flow velocity. A flow factor is provided to convert frequency to volumetric flow rate for each size of flowmeter. Flowmeters have a PT1000 RTD for temperature measurement of the media
Note: Media with viscosity greater than 2 centipose will require higher flow rates to form measurable vortices within the flowtube.
5 Options Available Below
Product Details & Documents
Built-in 1000 Ω RTD sensor provides direct temperature measurement of media.
- Deigned with rugged PPA molded construction for enhanced durability
- Excellent media compatibility with ETFE, EPDM, and PPD wetted parts
- Use with low viscosity liquids for laboratory and industrial applications
These economical vortex flowmeters are suitable for water and water glycol-based heat exchange systems. The transmitters work on the vortex shedding flow principle. As flow velocity increases, alternating vortices are formed on each side of the bluff body within the flowtube and travel downstream. The flowmeters utilize piezoelectric sensors embedded in a vane downstream from the bluff to detect the vortices and transfer the sensed vortices into flow velocity. A flow factor is provided to convert frequency to volumetric flow rate for each size of flowmeter. Flowmeters have a PT1000 RTD for temperature measurement of the media
Note: Media with viscosity greater than 2 centipose will require higher flow rates to form measurable vortices within the flowtube.
Specifications for Product Family
Especificaciones
- Accuracy:
- Up to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of reading
- Up to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of reading
- Up to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of reading
- Up to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of reading
- Up to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of reading
- Connection:
- M12×1, 5-pole DC microconnector
- M12×1, 5-pole DC microconnector
- M12×1, 5-pole DC microconnector
- M12×1, 5-pole DC microconnector
- M12×1, 5-pole DC microconnector
- Min. operating temperature:
- –15 °C (5 °F)
- –15 °C (5 °F)
- –15 °C (5 °F)
- –15 °C (5 °F)
- –15 °C (5 °F)
- Max. operating temperature:
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- Max. pressure:
- 261 psi
- 261 psi
- 261 psi
- 261 psi
- 261 psi
- Pressure drop at max. flow:
- 3 psid
- 3 psid
- 3 psid
- 3 psid
- 3 psid
- Height:
- 59710"
- 59710"
- 59710"
- 59710"
- 59710"
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Especificaciones
Product Family Options
Información del producto
- Process ConnectionMin. flow rateMaximum Flow RateOutputAvailabilityPrice
- MFLX32825-68Vortex Flowmeter for Liquids, 0,24 to 3,96 gal./min / 0,9 to 15 L/minVortex Flowmeter for Liquids, 0,24 to 3,96 gal./min / 0,9 to 15 L/min0.25" NPT(F)0.25 gal./min3.96 gal./h4 to 20 mALog in Para disponibilidadEach (1UN)$6,070.00Especificaciones:IP65 washdown
More
Specifications
Cat. No.MFLX32825-68AccuracyUp to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of readingConnectionM12×1, 5-pole DC microconnectorMin. operating temperature–15 °C (5 °F)Max. operating temperature85 °C (185 °F)Max. pressure261 psiPressure drop at max. flow3 psidHeight59710"Warranty1 yearIP65 washdown - MFLX32825-70Vortex Flowmeter for Liquids, 0,48 to 8,45 gal./min / 1,8 to 32 L/minVortex Flowmeter for Liquids, 0,48 to 8,45 gal./min / 1,8 to 32 L/min0.375" NPT(F)0.48 gal./min8.45 gal./h4 to 20 mALog in Para disponibilidadEach (1UN)$6,360.00Especificaciones:IP65 washdown
More
Specifications
Cat. No.MFLX32825-70AccuracyUp to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of readingConnectionM12×1, 5-pole DC microconnectorMin. operating temperature–15 °C (5 °F)Max. operating temperature85 °C (185 °F)Max. pressure261 psiPressure drop at max. flow3 psidHeight59710"Warranty1 yearIP65 washdown - MFLX32825-72Vortex Flowmeter for Liquids, 0,93 to 13,2 gal./min / 3,5 to 50 L/minVortex Flowmeter for Liquids, 0,93 to 13,2 gal./min / 3,5 to 50 L/min0.5" NPT(F)39.6 gal./min13.2 gal./h4 to 20 mALog in Para disponibilidadEach (1UN)$6,420.00Especificaciones:IP65 washdown
More
Specifications
Cat. No.MFLX32825-72AccuracyUp to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of readingConnectionM12×1, 5-pole DC microconnectorMin. operating temperature–15 °C (5 °F)Max. operating temperature85 °C (185 °F)Max. pressure261 psiPressure drop at max. flow3 psidHeight59710"Warranty1 yearIP65 washdown - MFLX32825-74Vortex Flowmeter for Liquids, 1,32 to 22,5 gal./min / 5 to 85 L/minVortex Flowmeter for Liquids, 1,32 to 22,5 gal./min / 5 to 85 L/min0.75" NPT(F)1.32 gal./min22.5 gal./h4 to 20 mALog in Para disponibilidadEach (1UN)$7,010.00Especificaciones:IP65 washdown
More
Specifications
Cat. No.MFLX32825-74AccuracyUp to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of readingConnectionM12×1, 5-pole DC microconnectorMin. operating temperature–15 °C (5 °F)Max. operating temperature85 °C (185 °F)Max. pressure261 psiPressure drop at max. flow3 psidHeight59710"Warranty1 yearIP65 washdown - MFLX32825-76Vortex Flowmeter for Liquids, 2,38 to 39,6 gal./min / 9 to 150 L/minVortex Flowmeter for Liquids, 2,38 to 39,6 gal./min / 9 to 150 L/min1" NPT(F)2.38 gal./min39.6 gal./h4 to 20 mALog in Para disponibilidadEach (1UN)$6,460.00Especificaciones:IP65 washdown
More
Specifications
Cat. No.MFLX32825-76AccuracyUp to 50% of max flow: ±1% of reading; over 50% of max flow: ±2% of readingConnectionM12×1, 5-pole DC microconnectorMin. operating temperature–15 °C (5 °F)Max. operating temperature85 °C (185 °F)Max. pressure261 psiPressure drop at max. flow3 psidHeight59710"Warranty1 yearIP65 washdown