Orders LinkContactUsLinkComponent
Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
Supplier:   VWR International
Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
15 Options Available Below
Product Details & Documents
Measure 130 standard gases—user selectable from display.
  • Accuracy of ±0.8% of reading ±0.2% full-scale; repeatability of ±0.2%
  • Measure 30 standard gases—user selectable from display
  • 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM
  • Models with 4 to 20 mA output or totalizer are available
These meters measure flow via pressure drop across a laminar flow element (LFE). Because the flow element makes the flow stream laminar, placement in the process does not require straight pipe runs upstream or downstream of the meter, greatly simplifying installation. As compared to thermal mass technologies, the LFE design provides an ultrafast response within 10 milliseconds and offers “instant on” with no warm-up time.
An integrated keypad around the display is all that is required to program the unit for service. The 0 to 5 VDC output allows transmission of the flow value to a remote display, recorder, or controller regulating a valve or pump.
Flow controllers feature an integrated PID to direct the unit’s response to process changes. Flow set point is established with keypad, the optional set point control module, a 0 to 5 V signal, or an RS-232 input signal. Order set point control modules separately from the table. For portable flow metering applications, order the battery pack (sold separately).
Delivery information: Includes 120/230 VAC power adapter with communications cable and NIST-traceable calibration report supplied by the manufacturer.
Specifications for Product Family
Specifications
  • Certification:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Min. operating temperature:
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • –10 °C (14 °F)
  • Response time:
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • 50 ms
  • Model:
  • 32907-51
  • 32907-53
  • 32907-55
  • 32907-57
  • 32907-59
  • 32907-61
  • 32907-63
  • 32907-67
  • 32907-69
  • 32907-71
  • 32907-73
  • 32907-75
  • 32907-77
  • 32907-79
  • 32907-81
  • Min. flow rate:
  • 0.01 ml/min
  • 0.05 ml/min
  • 0.1 ml/min
  • 0.5 ml/min
  • 1 ml/min
  • 2 ml/min
  • 5 ml/min
  • 10 ml/min
  • 50 ml/min
  • 100 ml/min
  • 500 ml/min
  • 1000 ml/min
  • 2500 ml/min
  • 5000 ml/min
  • 10000 ml/min
  • Max. operating temperature:
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • 50 °C (122 °F)
  • Repeatability:
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.5%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • ±0.2%
  • Accuracy:
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • ±(0.8R + 0.2FS)
  • Output:
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • 0 to 5 VDC
  • Turndown ratio:
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • 100:1
  • Wetted materials:
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • 302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
  • Display type:
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • 7-line × 4-digit LCD; 0.25"H flow display
  • Communication interface:
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • Process connection:
  • 10-32 UNF
  • 10-32 UNF
  • 10-32 UNF
  • 10-32 UNF
  • ¹/₈" NPT(F)
  • ¹/₈" NPT(F)
  • ¹/₈" NPT(F)
  • ¹/₈" NPT(F)
  • ¹/₈" NPT(F)
  • ¹/₈" NPT(F)
  • ¹/₄" NPT(F)
  • ¹/₄" NPT(F)
  • ¹/₂" NPT(F)
  • ³/₄" NPT(F)
  • ³/₄" NPT(F)
  • Connection:
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • 8-pin circular mini-DIN
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Maximum Flow Rate
    Fluid Type
    Media Type
    Gas Type
    Availability
    Price
  • MFLX32907-51
    Gas Mass Flow Controller, 0.01 to 1.00 ml/min
    Gas Mass Flow Controller, 0.01 to 1.00 ml/min
    1 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $55,190.00
    $55,190.00
    Each (1UN)
       $55,190.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-51
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-51
    Min. flow rate
    0.01 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    10-32 UNF
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-53
    Gas Mass Flow Controller, 0.05 to 5.00 ml/min
    Gas Mass Flow Controller, 0.05 to 5.00 ml/min
    5 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $51,680.00
    $51,680.00
    Each (1UN)
       $51,680.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-53
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-53
    Min. flow rate
    0.05 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    10-32 UNF
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-55
    Gas Mass Flow Controller, 0.10 to 10.0 ml/min
    Gas Mass Flow Controller, 0.10 to 10.0 ml/min
    10 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $44,070.00
    $44,070.00
    Each (1UN)
       $44,070.00
    Specifications:
    CE Compliant
    CSA Certified
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-55
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-55
    Min. flow rate
    0.1 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    10-32 UNF
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    CSA Certified
    NIST traceable
  • MFLX32907-57
    Gas Mass Flow Controller, 0.50 to 50.0 ml/min
    Gas Mass Flow Controller, 0.50 to 50.0 ml/min
    50 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $44,070.00
    $44,070.00
    Each (1UN)
       $44,070.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-57
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-57
    Min. flow rate
    0.5 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    10-32 UNF
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-59
    Gas Mass Flow Controller, 1.00 to 100 ml/min
    Gas Mass Flow Controller, 1.00 to 100 ml/min
    100 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $39,780.00
    $39,780.00
    Each (1UN)
       $39,780.00
    Specifications:
    CE Compliant
    CSA Certified
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-59
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-59
    Min. flow rate
    1 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₈" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    CSA Certified
    NIST traceable
  • MFLX32907-61
    Gas Mass Flow Controller, 2.00 to 200 ml/min
    Gas Mass Flow Controller, 2.00 to 200 ml/min
    200 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $39,000.00
    $39,000.00
    Each (1UN)
       $39,000.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-61
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-61
    Min. flow rate
    2 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.5%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₈" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-63
    Gas Mass Flow Controller, 5.00 to 500 ml/min
    Gas Mass Flow Controller, 5.00 to 500 ml/min
    500 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $39,000.00
    $39,000.00
    Each (1UN)
       $39,000.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-63
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-63
    Min. flow rate
    5 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₈" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-67
    Gas Mass Flow Controller, 0.01 to 1 L/min
    Gas Mass Flow Controller, 0.01 to 1 L/min
    1000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $39,000.00
    $39,000.00
    Each (1UN)
       $39,000.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-67
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-67
    Min. flow rate
    10 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₈" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-69
    Gas Mass Flow Controller, 0.05 to 5 L/min
    Gas Mass Flow Controller, 0.05 to 5 L/min
    5000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $39,000.00
    $39,000.00
    Each (1UN)
       $39,000.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-69
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-69
    Min. flow rate
    50 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₈" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-71
    Gas Mass Flow Controller, for gas, 0.1 to 10 L/min
    Gas Mass Flow Controller, for gas, 0.1 to 10 L/min
    10000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $39,000.00
    $39,000.00
    Each (1UN)
       $39,000.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-71
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-71
    Min. flow rate
    100 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₈" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-73
    Gas Mass Flow Controller, 0.5 to 50 L/min
    Gas Mass Flow Controller, 0.5 to 50 L/min
    50000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $50,900.00
    $50,900.00
    Each (1UN)
       $50,900.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-73
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-73
    Min. flow rate
    500 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₄" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-75
    Gas Mass Flow Controller, 1 to 100 L/min
    Gas Mass Flow Controller, 1 to 100 L/min
    100000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $54,410.00
    $54,410.00
    Each (1UN)
       $54,410.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-75
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-75
    Min. flow rate
    1000 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₄" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-77
    Gas Mass Flow Controller, 2.500 to 250.0 L/min
    Gas Mass Flow Controller, 2.500 to 250.0 L/min
    250000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $59,090.00
    $59,090.00
    Each (1UN)
       $59,090.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-77
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-77
    Min. flow rate
    2500 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ¹/₂" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-79
    Gas Mass Flow Controller, 5 to 500 L/min
    Gas Mass Flow Controller, 5 to 500 L/min
    500000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $66,690.00
    $66,690.00
    Each (1UN)
       $66,690.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-79
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-79
    Min. flow rate
    5000 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ³/₄" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable
  • MFLX32907-81
    Gas Mass Flow Controller, 10 to 1000 L/min
    Gas Mass Flow Controller, 10 to 1000 L/min
    1000000 ml/min
    Gas
    Acetylene;Air;Argon;Carbon Dioxide;Carbon Monoxide;Helium;Hydrogen;Nitrogen;Oxygen
    Acetylene (C₂H₂), Air, Argon (Ar), i-Butane (i-C₄H₁₀), n-Butane (n-C₄H₁₀), Carbon Dioxide (CO₂), Carbon Monoxide (CO), Deuterium (D₂), Ethane (C₂H₆), Ethylene/Ethene (C₂H₄), Helium (He), Hydrogen (H₂), Krypton (Kr), Methane (CH₄), Neon (Ne), Nitrogen (N₂), Nitrous Oxide (N₂O), Oxygen (O₂), Propane (C₃H₈), Sulfur Hexaflouride (SF₆), Xenon (Xe)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $74,690.00
    $74,690.00
    Each (1UN)
       $74,690.00
    Specifications:
    CE Compliant
    NIST traceable
    More
    Specifications
    Certification
    Yes
    Cat. No.
    MFLX32907-81
    Min. operating temperature
    –10 °C (14 °F)
    Response time
    50 ms
    Model
    32907-81
    Min. flow rate
    10000 ml/min
    Max. operating temperature
    50 °C (122 °F)
    Repeatability
    ±0.2%
    Accuracy
    ±(0.8R + 0.2FS)
    Output
    0 to 5 VDC
    Turndown ratio
    100:1
    Wetted materials
    302, 303, and 410 SS; Viton®, silicone RTV, glass-reinforced nylon, aluminum, brass, silicon, glass
    Display type
    7-line × 4-digit LCD; 0.25"H flow display
    Communication interface
    RS-232
    Process connection
    ³/₄" NPT(F)
    Connection
    8-pin circular mini-DIN
    Warranty
    1 year
    CE Compliant
    NIST traceable