Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
Create your own flowmeter systems for complete flexibility.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
- Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
- Use with media for laboratory and industrial applications
- Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
- Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
- Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.
Create your own flowmeter systems for complete flexibility.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
- Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
- Use with media for laboratory and industrial applications
- Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
- Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
- Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.
- Max. operating temperature:
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- Max. pressure:
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Product Family Options
- DescriptionProcess ConnectionValveHousing Material
- Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Aluminum0.125" NPT(F)NoAluminiumEach (1UN)$1,300.00
Specifications
Cat. No.MFLX03220-00Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year - Variable-Area Single-Tube Frame, 150-mm Scale; No Valve Port, Aluminum0.125" NPT(F)NoAluminiumEach (1UN)$1,920.00
Specifications
Cat. No.MFLX03220-06Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year - Variable-Area Single-Tube Frame, 65-mm Scale; Valve Port, Aluminum0.125" NPT(F)YesAluminiumEach (1UN)$1,140.00
Specifications
Cat. No.MFLX03220-08Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year - Variable-Area Single-Tube Frame, 150-mm Scale; Valve Port, Aluminum0.125" NPT(F)YesAluminiumEach (1UN)$1,920.00
Specifications
Cat. No.MFLX03220-10Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year - Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Stainless Steel0.125" NPT(F)NoStainless steelEach (1UN)$3,550.00
Specifications
Cat. No.MFLX03220-40Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year - Variable-Area Single-Tube Frame, 65-mm Scale; Valve Port, Stainless Steel0.125" NPT(F)YesStainless steelEach (1UN)$3,550.00
Specifications
Cat. No.MFLX03220-44Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year - Variable-Area Single-Tube Frame, 150-mm Scale; Valve Port, Stainless Steel0.125" NPT(F)YesStainless steelEach (1UN)$4,200.00
Specifications
Cat. No.MFLX03220-46Max. operating temperature121 °C (250 °F)Max. pressure200 psiWarranty1 year