Orders LinkContactUsLinkComponent
FMI Temperature-Controlled Piston Pump Heads
FMI Temperature-Controlled Piston Pump Heads
Supplier:   FLUID METERING INC
FMI Temperature-Controlled Piston Pump Heads
Supplier:  FLUID METERING INC
Certificates
FMI Temperature-Controlled Piston Pump Heads
6 Options Available Below
Product Details & Documents
Pump viscous fluids or maintain process fluid temperatures with temperature-controlled pump head.
  • Operating temperature up to 350 °F
  • Handle pressures up to 100 psig
  • Wetted materials of ceramic, PTFE, and 316 SS
Suitable for applications to maintain process fluid temperatures or pump viscous solutions (such as waxes, pastes or cosmetics). Run wash fluids, gases or steam through gland ports to provide a barrier between process fluids and the seal area (atmosphere). Using this barrier maintains temperatures and pressures required of slurries, particulates, crystal formers and anaerobics.
Control and monitor the temperature of the pump head with a temperature panel meter (order power cord also), 2 standard cartridge heaters and thermocouple (order separately under accessories). To provide power to the pump head, order a drive under accessories.
Specifications for Product Family
Especificaciones
  • Model:
  • 07107-00
  • 07107-05
  • 07107-04
  • 07107-09
  • 07107-02
  • 07107-07
  • Accuracy:
  • ±0.1%
  • ±0.1%
  • ±0.1%
  • ±0.1%
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 0.25 NPT(F)
  • 0.25 NPT(F)
  • Max. pressure:
  • 100 psi
  • 50 psi
  • 60 psi
  • 60 psi
  • 60 psi
  • 60 psi
  • Max. temperature:
  • 176 °C (350 °F)
  • 176 °C (350 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Max. viscosity:
  • 500 cP
  • 500 cP
  • 500 cP
  • 500 cP
  • Min. flow rate:
  • 0 ml/min
  • 0 ml/min
  • Outlet connection:
  • 0.25 NPT(F)
  • 0.25 NPT(F)
  • Power connection:
  • ¹/₈" NPT(F)
  • ¹/₈" NPT(F)
  • Suction lift, wet:
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • Wetted materials:
  • 316 SS, ceramic, PTFE
  • 316 SS, ceramic, PTFE
  • Kynar® cylinder, ceramic diaphragm & cylinder liner
  • Kynar® cylinder, ceramic diaphragm & cylinder liner
  • Kynar® cylinder, ceramic diaphragm & cylinder liner
  • Kynar® cylinder, ceramic diaphragm & cylinder liner
Especificaciones
Product Family Options
Información del producto
  • Maximum Flow Rate
    Ø
    Availability
    Price
  • MFLX07107-00
    Temperature-Controlled Pump Head, 0 to 576 ml/min at 1800 rpm
    Temperature-Controlled Pump Head, 0 to 576 ml/min at 1800 rpm
    576 ml/min
    6.35 mm (0.25")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $19,290.00
    $19,290.00
    Each (1UN)
       $19,290.00
    Supplier Number: Q1CSCWT
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07107-00
    Model
    07107-00
    Accuracy
    Duty cycle continuous
    Inlet connection
    0.25 NPT(F)
    Max. pressure
    100 psi
    Max. temperature
    176 °C (350 °F)
    Max. viscosity
    Min. flow rate
    0 ml/min
    Outlet connection
    0.25 NPT(F)
    Power connection
    ¹/₈" NPT(F)
    Suction lift, wet
    Wetted materials
    316 SS, ceramic, PTFE
  • MFLX07107-05
    Temperature-Controlled Pump Head, 0 to 1296 ml/min at 1800 rpm
    Temperature-Controlled Pump Head, 0 to 1296 ml/min at 1800 rpm
    1296 ml/min
    9.53 mm (0.375")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18,780.00
    $18,780.00
    Each (1UN)
       $18,780.00
    Supplier Number: Q2CSCWT
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07107-05
    Model
    07107-05
    Accuracy
    Duty cycle continuous
    Inlet connection
    0.25 NPT(F)
    Max. pressure
    50 psi
    Max. temperature
    176 °C (350 °F)
    Max. viscosity
    Min. flow rate
    0 ml/min
    Outlet connection
    0.25 NPT(F)
    Power connection
    ¹/₈" NPT(F)
    Suction lift, wet
    Wetted materials
    316 SS, ceramic, PTFE
  • MFLX07107-04
    Valveless Piston Pump Head, Ceramic/PVDF/Ceramic with Isolation Gland; 0,016 ml/rev
    Valveless Piston Pump Head, Ceramic/PVDF/Ceramic with Isolation Gland; 0,016 ml/rev
    192 ml/min
    6.35 mm (0.25")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,130.00
    $13,130.00
    Each (1UN)
       $13,130.00
    Supplier Number: Q1CKCW
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07107-04
    Model
    07107-04
    Accuracy
    ±0.1%
    Duty cycle continuous
    Yes
    Inlet connection
    Max. pressure
    60 psi
    Max. temperature
    100 °C (212 °F)
    Max. viscosity
    500 cP
    Min. flow rate
    Outlet connection
    Power connection
    Suction lift, wet
    4572 mm of H₂O (180" H₂O)
    Wetted materials
    Kynar® cylinder, ceramic diaphragm & cylinder liner
  • MFLX07107-09
    Valveless Piston Pump Head, Ceramic/PVDF/Ceramic with Isolation Gland; 0,036 ml/rev
    Valveless Piston Pump Head, Ceramic/PVDF/Ceramic with Isolation Gland; 0,036 ml/rev
    192 ml/min
    6.35 mm (0.25")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,130.00
    $13,130.00
    Each (1UN)
       $13,130.00
    Supplier Number: Q2CKCW
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07107-09
    Model
    07107-09
    Accuracy
    ±0.1%
    Duty cycle continuous
    Yes
    Inlet connection
    Max. pressure
    60 psi
    Max. temperature
    100 °C (212 °F)
    Max. viscosity
    500 cP
    Min. flow rate
    Outlet connection
    Power connection
    Suction lift, wet
    4572 mm of H₂O (180" H₂O)
    Wetted materials
    Kynar® cylinder, ceramic diaphragm & cylinder liner
  • MFLX07107-02
    Valveless Piston Pump Head, Ceramic/Stainless Steel/Ceramic with Isolation Gland; 0,016 ml/rev
    Valveless Piston Pump Head, Ceramic/Stainless Steel/Ceramic with Isolation Gland; 0,016 ml/rev
    192 ml/min
    6.35 mm (0.25")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $19,890.00
    $19,890.00
    Each (1UN)
       $19,890.00
    Supplier Number: Q1CSCW
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07107-02
    Model
    07107-02
    Accuracy
    ±0.1%
    Duty cycle continuous
    Yes
    Inlet connection
    Max. pressure
    60 psi
    Max. temperature
    100 °C (212 °F)
    Max. viscosity
    500 cP
    Min. flow rate
    Outlet connection
    Power connection
    Suction lift, wet
    4572 mm of H₂O (180" H₂O)
    Wetted materials
    Kynar® cylinder, ceramic diaphragm & cylinder liner
  • MFLX07107-07
    Valveless Piston Pump Head, Ceramic/Stainless Steel/Ceramic with Isolation Gland; 0,036 ml/rev
    Valveless Piston Pump Head, Ceramic/Stainless Steel/Ceramic with Isolation Gland; 0,036 ml/rev
    192 ml/min
    6.35 mm (0.25")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18,570.00
    $18,570.00
    Each (1UN)
       $18,570.00
    Supplier Number: Q2CSCW
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07107-07
    Model
    07107-07
    Accuracy
    ±0.1%
    Duty cycle continuous
    Yes
    Inlet connection
    Max. pressure
    60 psi
    Max. temperature
    100 °C (212 °F)
    Max. viscosity
    500 cP
    Min. flow rate
    Outlet connection
    Power connection
    Suction lift, wet
    4572 mm of H₂O (180" H₂O)
    Wetted materials
    Kynar® cylinder, ceramic diaphragm & cylinder liner
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Series Q pump head to drive adapter kit
    Series Q pump head to drive adapter kit
    MFLX07104-48
    Log in Para disponibilidad
    Each (1UN) - $18,410.00