Masterflex® L/S® Cytoflow™ Pump Heads, Avantor®
Acerca de este artículo
The best option for pumping live cells and shear-sensitive fluids—validated by independent test data.
- Convex-roller design is gentler on live cells and shear-sensitive fluids—validated by independent test data
- Large-diameter rotor delivers higher flow rates at lower speeds
- Ensure the survival and vitality of live cells
- Flow rate 0.03 to 5700 ml/min with L/S tubing—flow rate depends on the drive rpm, pump head, and tubing size
- Compatible with all Masterflex L/S drives that accept two or more pump heads
- Models available for Masterflex L/S Precision and L/S High-Performance precision pump tubing
- 2-roller and 3-roller pump heads available--choose 2-roller for maximum flow, 3-roller for less pulsation
Developed specifically for pumping delicate live cells and shear-sensitive fluids, the Masterflex L/S Cytoflow pump head is ideal for use in biopharma and microbiology applications where peristaltic pump operators want to minimize cell hemolysis. The extra-large tubing bed combined with fewer rollers results in high flow rates at low motor speeds, which furthers its cell viability advantages.
Optimum rpm (drive motor speed) range for transferring live cells is <100 rpm; optimum rpm for other shear-sensitive fluids is <300 rpm; optimum rpm for all other general pumping applications is up to 600 rpm.
4 Options Available Below
Product Details & Documents
The best option for pumping live cells and shear-sensitive fluids—validated by independent test data.
- Convex-roller design is gentler on live cells and shear-sensitive fluids—validated by independent test data
- Large-diameter rotor delivers higher flow rates at lower speeds
- Ensure the survival and vitality of live cells
- Flow rate 0.03 to 5700 ml/min with L/S tubing—flow rate depends on the drive rpm, pump head, and tubing size
- Compatible with all Masterflex L/S drives that accept two or more pump heads
- Models available for Masterflex L/S Precision and L/S High-Performance precision pump tubing
- 2-roller and 3-roller pump heads available--choose 2-roller for maximum flow, 3-roller for less pulsation
Developed specifically for pumping delicate live cells and shear-sensitive fluids, the Masterflex L/S Cytoflow pump head is ideal for use in biopharma and microbiology applications where peristaltic pump operators want to minimize cell hemolysis. The extra-large tubing bed combined with fewer rollers results in high flow rates at low motor speeds, which furthers its cell viability advantages.
Optimum rpm (drive motor speed) range for transferring live cells is <100 rpm; optimum rpm for other shear-sensitive fluids is <300 rpm; optimum rpm for all other general pumping applications is up to 600 rpm.
Specifications for Product Family
Especificaciones
- Model:
- 77253-00
- 77253-02
- 77253-10
- 77253-12
- Gear ratio:
- 1:1
- 1:1
- 1:1
- 1:1
- No. of channels:
- 1
- 1
- 1
- 1
- Occlusion:
- Fixed
- Fixed
- Fixed
- Fixed
- Open-head sensor:
- No
- No
- No
- No
- Optimum RPM range:
- 0.02 - 600 rpm
- 0.02 - 600 rpm
- 0.02 - 600 rpm
- 0.02 - 600 rpm
- Pump series:
- L/S
- L/S
- L/S
- L/S
- Tubing retention:
- Manual
- Manual
- Manual
- Manual
- Tubing sizes accepted:
- L/S® 16, L/S 25, L/S 17, L/S 18
- L/S® 15, L/S 24, L/S 35, L/S 36
- L/S® 16, L/S 25, L/S 17, L/S 18
- L/S® 15, L/S 24, L/S 35, L/S 36
- Volume per revolution with L/S 15 tubing:
- 4 ml
- 3.5 ml
- Volume per revolution with L/S 16 tubing:
- 1.7 ml
- 1.6 ml
- Volume per revolution with L/S 17 tubing:
- 6.1 ml
- 5.6 ml
- Volume per revolution with L/S 18 tubing:
- 8.6 ml
- 7.7 ml
- Volume per revolution with L/S 24 tubing:
- 6.2 ml
- 5.7 ml
- Volume per revolution with L/S 25 tubing:
- 3.9 ml
- 3.5 ml
- Volume per revolution with L/S 35 tubing:
- 9.5 ml
- 7.8 ml
- Volume per revolution with L/S 36 tubing:
- 12.8 ml
- 9.6 ml
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
Especificaciones
Product Family Options
Información del producto
- RollersMaximum Flow RateHousing MaterialRotor MaterialHeightLengthWidthAvailabilityPrice
- MFLX77253-00L/S® Cytoflow™ Pump Head for Precision Tubing, 2-RollerL/S® Cytoflow™ Pump Head for Precision Tubing, 2-Roller25100 ml/minAnodized aluminiumAluminum115 mm (4.5")89 mm (3.5")115 mm (4.5")Log in Para disponibilidadEach (1UN)$28,670.00Supplier Number: 77253-00Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77253-00Model77253-00Gear ratio1:1No. of channels1OcclusionFixedOpen-head sensorNoOptimum RPM range0.02 - 600 rpmPump seriesL/STubing retentionManualTubing sizes acceptedL/S® 16, L/S 25, L/S 17, L/S 18Volume per revolution with L/S 15 tubingVolume per revolution with L/S 16 tubing1.7 mlVolume per revolution with L/S 17 tubing6.1 mlVolume per revolution with L/S 18 tubing8.6 mlVolume per revolution with L/S 24 tubingVolume per revolution with L/S 25 tubing3.9 mlVolume per revolution with L/S 35 tubingVolume per revolution with L/S 36 tubingWarranty2 years2 year warranty - MFLX77253-02L/S® Cytoflow™ 2- Roller low shear pump head, 7600 ml/min max flowL/S® Cytoflow™ 2- Roller low shear pump head, 7600 ml/min max flow27600 ml/minAnodized aluminiumAluminum115 mm (4.5")89 mm (3.5")115 mm (4.5")Log in Para disponibilidadEach (1UN)$28,670.00Supplier Number: 77253-02Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77253-02Model77253-02Gear ratio1:1No. of channels1OcclusionFixedOpen-head sensorNoOptimum RPM range0.02 - 600 rpmPump seriesL/STubing retentionManualTubing sizes acceptedL/S® 15, L/S 24, L/S 35, L/S 36Volume per revolution with L/S 15 tubing4 mlVolume per revolution with L/S 16 tubingVolume per revolution with L/S 17 tubingVolume per revolution with L/S 18 tubingVolume per revolution with L/S 24 tubing6.2 mlVolume per revolution with L/S 25 tubingVolume per revolution with L/S 35 tubing9.5 mlVolume per revolution with L/S 36 tubing12.8 mlWarranty2 years2 year warranty - MFLX77253-10L/S® Cytoflow™ Pump Head for Precision Tubing, 3-RollerL/S® Cytoflow™ Pump Head for Precision Tubing, 3-Roller34600 ml/minAnodized aluminiumAluminum115 mm (4.5")89 mm (3.5")115 mm (4.5")Log in Para disponibilidadEach (1UN)$29,840.00Supplier Number: 77253-10Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77253-10Model77253-10Gear ratio1:1No. of channels1OcclusionFixedOpen-head sensorNoOptimum RPM range0.02 - 600 rpmPump seriesL/STubing retentionManualTubing sizes acceptedL/S® 16, L/S 25, L/S 17, L/S 18Volume per revolution with L/S 15 tubingVolume per revolution with L/S 16 tubing1.6 mlVolume per revolution with L/S 17 tubing5.6 mlVolume per revolution with L/S 18 tubing7.7 mlVolume per revolution with L/S 24 tubingVolume per revolution with L/S 25 tubing3.5 mlVolume per revolution with L/S 35 tubingVolume per revolution with L/S 36 tubingWarranty2 years2 year warranty - MFLX77253-12L/S® Cytoflow™ Pump Head for High-Performance Precision Tubing, 3-RollerL/S® Cytoflow™ Pump Head for High-Performance Precision Tubing, 3-Roller35700 ml/minAnodized aluminiumAluminum115 mm (4.5")89 mm (3.5")115 mm (4.5")Log in Para disponibilidadEach (1UN)$29,840.00Supplier Number: 77253-12Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77253-12Model77253-12Gear ratio1:1No. of channels1OcclusionFixedOpen-head sensorNoOptimum RPM range0.02 - 600 rpmPump seriesL/STubing retentionManualTubing sizes acceptedL/S® 15, L/S 24, L/S 35, L/S 36Volume per revolution with L/S 15 tubing3.5 mlVolume per revolution with L/S 16 tubingVolume per revolution with L/S 17 tubingVolume per revolution with L/S 18 tubingVolume per revolution with L/S 24 tubing5.7 mlVolume per revolution with L/S 25 tubingVolume per revolution with L/S 35 tubing7.8 mlVolume per revolution with L/S 36 tubing9.6 mlWarranty2 years2 year warranty