Masterflex® B/T® Rapid-Load Pump Heads, Avantor®
About this item
Convert your existing motor into a high-capacity Masterflex® B/T® drive.
- Newly designed occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
- Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
- Interlock switch shuts off power to stop drive when door is opened
- Three rollers and 5.6:1 hear ratio provides optimal flow capacity
- Nylatron® rotor / roller for greater durability and chemical resistance
Enjoy all the advantages of peristaltic tubing drives using your existing motor. Gentle pumping action is ideal for viscous or shear-sensitive fluids, including live cells and cell products, slurries, and suspensions. Easy clean-up and change-over between batches—simply throw the tubing away and install a new piece in seconds. Tubing is the only wetted part so chemical compatibility is greatly simplified, while maintenance is quick and easy.
B/T Rapid-Load pump heads ensure fast and easy tubing changes and feature an occlusion mechanism that slides the tubing occlusion bed out of the way for easy and repeatable tube loading. Pump head door is constructed of powder-coated cast aluminum and polycarbonate, frame is made of durable stainless steel/powder-coated carbon steel, and rollers are Nylatron® for rugged dependability. IP56-rated housing protects against dust and water.
2 Options Available Below
Product Details & Documents
Convert your existing motor into a high-capacity Masterflex® B/T® drive.
- Newly designed occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
- Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
- Interlock switch shuts off power to stop drive when door is opened
- Three rollers and 5.6:1 hear ratio provides optimal flow capacity
- Nylatron® rotor / roller for greater durability and chemical resistance
Enjoy all the advantages of peristaltic tubing drives using your existing motor. Gentle pumping action is ideal for viscous or shear-sensitive fluids, including live cells and cell products, slurries, and suspensions. Easy clean-up and change-over between batches—simply throw the tubing away and install a new piece in seconds. Tubing is the only wetted part so chemical compatibility is greatly simplified, while maintenance is quick and easy.
B/T Rapid-Load pump heads ensure fast and easy tubing changes and feature an occlusion mechanism that slides the tubing occlusion bed out of the way for easy and repeatable tube loading. Pump head door is constructed of powder-coated cast aluminum and polycarbonate, frame is made of durable stainless steel/powder-coated carbon steel, and rollers are Nylatron® for rugged dependability. IP56-rated housing protects against dust and water.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX77111-50
- MFLX77111-55
- Model:
- 77111-50
- 77111-55
- Gear ratio:
- 5.6:1
- 5.6:1
- IP rating:
- IP 56
- IP 56
- Motor mount:
- NEMA Type 56C
- IEC 72-71-14F130 with B5 160-mm flange
- Mounting type:
- Direct-coupled
- Direct-coupled
- No. of rollers:
- 3
- 3
- Open-head sensor:
- No
- No
- Pump series:
- B/T
- B/T
- Tubing sizes accepted:
- B/T 87 or B/T 91
- B/T 87 or B/T 91
- Volume per revolution with B/T 87 tubing:
- 0.059 L (0.0156 gal.)
- 0.059 L (0.0156 gal.)
- Volume per revolution with B/T 91 tubing:
- 0.131 L (0.035 gal.)
- 0.131 L (0.035 gal.)
- Warranty:
- 2 years
- 2 years
Specifications
Product Family Options
Product Information
- Maximum Flow RateHeightLengthWidthAvailabilityPrice
- MFLX77111-50B/T® Rapid-Load® Pump Head for NEMA Type 56C MotorB/T® Rapid-Load® Pump Head for NEMA Type 56C Motor42000 ml/min (11.1 gal./min)400 mm (15.7968")384 mm (15.1093")686 mm (27")Log in for availabilityEach (1UN)$126,750.00Supplier Number: 77111-50Specifications:2 year warrantyIP56 washdown
More
Specifications
Cat. No.MFLX77111-50Model77111-50Gear ratio5.6:1IP ratingIP 56Motor mountNEMA Type 56CMounting typeDirect-coupledNo. of rollers3Open-head sensorNoPump seriesB/TTubing sizes acceptedB/T 87 or B/T 91Volume per revolution with B/T 87 tubing0.059 L (0.0156 gal.)Volume per revolution with B/T 91 tubing0.131 L (0.035 gal.)Warranty2 years2 year warrantyIP56 washdown - MFLX77111-55B/T® Rapid-Load® Pump Head for IEC 72-71-14F130 MotorB/T® Rapid-Load® Pump Head for IEC 72-71-14F130 Motor42000 ml/min (11.1 gal./min)400 mm (15.7968")384 mm (15.1093")686 mm (27")Log in for availabilityEach (1UN)$128,510.00Supplier Number: 77111-55Specifications:2 year warrantyCE CompliantIP56 washdown
More
Specifications
Cat. No.MFLX77111-55Model77111-55Gear ratio5.6:1IP ratingIP 56Motor mountIEC 72-71-14F130 with B5 160-mm flangeMounting typeDirect-coupledNo. of rollers3Open-head sensorNoPump seriesB/TTubing sizes acceptedB/T 87 or B/T 91Volume per revolution with B/T 87 tubing0.059 L (0.0156 gal.)Volume per revolution with B/T 91 tubing0.131 L (0.035 gal.)Warranty2 years2 year warrantyCE CompliantIP56 washdown