ContactUsLinkComponent
Masterflex® I/P® High-Performance Pump Head Replacement Parts, Avantor®
Masterflex® I/P® High-Performance Pump Head Replacement Rotor and Body Assembly
Catálogo # MFLX77600-69
undefined
Masterflex® I/P® High-Performance Pump Head Replacement Rotor and Body Assembly
Catálogo # MFLX77600-69
Número de proveedor:  77600-69
Especificaciones
  • Pump series:
    I/P
  • Descripción:
    KIT REPL ROTOR & BODY ASSEMBLY (Rotor/body assembly with tang boot)
  • Referencia:
    MFLX77600-69
  • Rotor Material:
    Stainless steel
  • Housing Material:
    Stainless steel, thermoset polyester
Especificaciones
Acerca de este artículo
Get the most our of your Masterflex® I/P® High-Performance Pump Head with these replacement parts.
  • Stainless steel (SS) rotor / roller for greater durability and chemical resistance
Replacement parts to keep your pump head operating even longer.
Entrega: Includes rotor, body, and tang boot.
  • Pump series:
    I/P
  • Descripción:
    KIT REPL ROTOR & BODY ASSEMBLY (Rotor/body assembly with tang boot)
  • Environmentally Preferable:
  • Referencia:
    MFLX77600-69
  • Rotor Material:
    Stainless steel
  • Housing Material:
    Stainless steel, thermoset polyester