Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
Certificates
Acerca de este artículo
Compact, cost effective pump systems for general fluid transfer applications.
- Simple operation with single-turn speed control
- Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
- Reversible motor lets you easily prime/purge tubing and pump in either direction
- Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.
10 Options Available Below
Product Details & Documents
Compact, cost effective pump systems for general fluid transfer applications.
- Simple operation with single-turn speed control
- Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
- Reversible motor lets you easily prime/purge tubing and pump in either direction
- Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.
Specifications for Product Family
Especificaciones
- Catalog No:
- MFLX77910-40
- MFLX77910-45
- MFLX77910-50
- MFLX77910-55
- MFLX77910-60
- MFLX77910-65
- MFLX77910-70
- MFLX77910-75
- MFLX77715-90
- MFLX77715-91
- Model:
- 77910-40
- 77910-50
- 77910-50
- 77910-55
- 77910-60
- 77910-65
- 77910-70
- 77910-75
- No. of channels:
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- Channels:
- 1
- 1
- No. of rollers:
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4; stainless steel
- 4; stainless steel
- Control type:
- Variable-Speed Analog
- Variable-Speed Analog
- Analog
- Analog
- Analog
- Analog
- Analog
- Analog
- Housing material:
- Polyarylamide (PARA)
- Polyarylamide (PARA)
- Min. flow rate per channel:
- 16 ml/min
- 16 ml/min
- 56 ml/min
- 56 ml/min
- 16 ml/min
- 16 ml/min
- 56 ml/min
- 56 ml/min
- 0.06 ml/min
- 0.06 ml/min
- Max. flow rate per channel:
- 480 ml/min
- 480 ml/min
- 1700 ml/min
- 1700 ml/min
- 480 ml/min
- 480 ml/min
- 1700 ml/min
- 1700 ml/min
- 3.8 ml/min
- 3.8 ml/min
- Drive IP rating:
- IP 22
- IP 22
- IP 22
- IP 22
- IP 22
- IP 22
- IP 22
- IP 22
- Accuracy:
- ±5%
- ±5%
- ±5%
- ±5%
- ±5%
- ±5%
- ±5%
- ±5%
- Anti-drip:
- No
- No
- No
- No
- No
- No
- No
- No
- Cloud connected:
- No
- No
- No
- No
- No
- No
- No
- No
- Digital dispensing:
- No
- No
- No
- No
- No
- No
- No
- No
- Drive:
- 07555-00 (L/S® Analog Variable-Speed Console Drive)
- 07555-05 (L/S® Analog Variable-Speed Console Drive)
- 07555-00 (L/S® Analog Variable-Speed Console Drive)
- 07555-05 (L/S® Analog Variable-Speed Console Drive)
- 07555-00 (L/S® Analog Variable-Speed Console Drive)
- 07555-05 (L/S® Analog Variable-Speed Console Drive)
- 07555-00 (L/S® Analog Variable-Speed Console Drive)
- 07555-05 (L/S® Analog Variable-Speed Console Drive)
- Duty cycle:
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Input:
- Start/Stop (remote inputs)
- Start/Stop (remote inputs)
- Min. RPM:
- 20 rpm
- 20 rpm
- 20 rpm
- 20 rpm
- 20 rpm
- 20 rpm
- 20 rpm
- 20 rpm
- Open-head sensor:
- No
- No
- No
- No
- No
- No
- No
- No
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Remote I/O:
- Start/Stop
- Start/Stop
- Start/Stop
- Start/Stop
- Start/Stop
- Start/Stop
- Start/Stop
- Start/Stop
- Reversible motor:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Tubing:
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- Tubing sizes accepted:
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, L/S 24
- L/S® 15, 24
- Washdown capable:
- No
- No
- No
- No
- No
- No
- No
- No
- Current:
- 1.5 A
- 0.8 A
- 1.5 A
- 0.8 A
- 1.5 A
- 0.8 A
- 1.5 A
- 0.8 A
- Frequency:
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- Voltage:
- 115 VAC
- 230 VAC
- 115 VAC
- 230 VAC
- 115 VAC
- 230 VAC
- 115 VAC
- 230 VAC
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Especificaciones
Product Family Options
US plug models
Información del producto
- Plug TypeMaximum Flow RateHeightLengthWidthPump HeadPowerMax. RPMHousing MaterialAvailabilityPrice
- MFLX77910-40L/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for Precision Tubing; 115 VACL/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for Precision Tubing; 115 VACUS480 ml/min133.4 mm (5.25")256 mm (10.08")228.6 mm (9")77200-60 (L/S® Easy-Load® II head)115 VAC; 50/60 Hz; 1.5 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$41,540.00Supplier Number: 77910-40Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-40Model77910-40No. of channels1ChannelsNo. of rollers4Control typeVariable-Speed AnalogHousing materialMin. flow rate per channel16 ml/minMax. flow rate per channel480 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-00 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.5 AFrequency50/60 HzVoltage115 VACWarranty2 years2 year warranty - MFLX77910-45L/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for Precision Tubing; 230 VACL/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for Precision Tubing; 230 VACUS480 ml/min133.4 mm (5.25")256 mm (10.08")228.6 mm (9")77200-60 (L/S® Easy-Load® II head)230 VAC; 50/60 Hz; 0.8 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$41,730.00Supplier Number: 77910-45Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-45Model77910-50No. of channels1ChannelsNo. of rollers4Control typeVariable-Speed AnalogHousing materialMin. flow rate per channel16 ml/minMax. flow rate per channel480 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-05 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent0.8 AFrequency50/60 HzVoltage230 VACWarranty2 years2 year warranty - MFLX77910-50L/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for High-Performance Tubing; 115 VACL/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for High-Performance Tubing; 115 VACUS1700 ml/min133.4 mm (5.25")235.7 mm (9.28")228.6 mm (9")77200-62 (L/S® Easy-Load® II pump head)115 VAC; 50/60 Hz; 1.5 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$41,540.00Supplier Number: 77910-50Especificaciones:2 year warrantyCSA Certified
More
Specifications
Cat. No.MFLX77910-50Model77910-50No. of channels1ChannelsNo. of rollers4Control typeAnalogHousing materialMin. flow rate per channel56 ml/minMax. flow rate per channel1700 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-00 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.5 AFrequency50/60 HzVoltage115 VACWarranty2 years2 year warrantyCSA Certified - MFLX77910-55L/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for High-Performance Tubing; 230 VACL/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for High-Performance Tubing; 230 VACUS1700 ml/min133.4 mm (5.25")235.7 mm (9.28")228.6 mm (9")77200-62 (L/S® Easy-Load® II pump head)230 VAC; 50/60 Hz; 0.8 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$41,540.00Supplier Number: 77910-55Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-55Model77910-55No. of channels1ChannelsNo. of rollers4Control typeAnalogHousing materialMin. flow rate per channel56 ml/minMax. flow rate per channel1700 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-05 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent0.8 AFrequency50/60 HzVoltage230 VACWarranty2 years2 year warranty - MFLX77910-60L/S® Variable Speed Pump with Easy-Load® Precision Pump Head; 115 VACL/S® Variable Speed Pump with Easy-Load® Precision Pump Head; 115 VACUS480 ml/min133.4 mm (5.25")292 mm (11.5")184 mm (7.25")07514-10 (L/S® Easy-Load® head)115 VAC; 50/60 Hz; 1.5 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$39,390.00Supplier Number: 77910-60Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-60Model77910-60No. of channels1ChannelsNo. of rollers4Control typeAnalogHousing materialMin. flow rate per channel16 ml/minMax. flow rate per channel480 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-00 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.5 AFrequency50/60 HzVoltage115 VACWarranty2 years2 year warranty - MFLX77910-65L/S® Variable Speed Pump with Easy-Load® Precision Pump Head; 230 VACL/S® Variable Speed Pump with Easy-Load® Precision Pump Head; 230 VACUS480 ml/min133.4 mm (5.25")292 mm (11.5")184 mm (7.25")07514-10 (L/S® Easy-Load® head)230 VAC; 50/60 Hz; 0.8 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$39,590.00Supplier Number: 77910-65Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-65Model77910-65No. of channels1ChannelsNo. of rollers4Control typeAnalogHousing materialMin. flow rate per channel16 ml/minMax. flow rate per channel480 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-05 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent0.8 AFrequency50/60 HzVoltage230 VACWarranty2 years2 year warranty - MFLX77910-70L/S® Variable Speed Pump with Easy-Load® High-Performance Precision Pump Head; 115 VACL/S® Variable Speed Pump with Easy-Load® High-Performance Precision Pump Head; 115 VACUS1700 ml/min133.4 mm (5.25")292 mm (11.5")184 mm (7.25")07514-12 (L/S® Easy-Load® head)115 VAC; 50/60 Hz; 1.5 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$39,390.00Supplier Number: 77910-70Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-70Model77910-70No. of channels1ChannelsNo. of rollers4Control typeAnalogHousing materialMin. flow rate per channel56 ml/minMax. flow rate per channel1700 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-00 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)Tubing sizes acceptedL/S® 15, L/S 24Washdown capableNoCurrent1.5 AFrequency50/60 HzVoltage115 VACWarranty2 years2 year warranty - MFLX77910-75L/S® Variable Speed Pump with Easy-Load® High-Performance Precision Pump Head; 230 VACL/S® Variable Speed Pump with Easy-Load® High-Performance Precision Pump Head; 230 VACUS1700 ml/min133.4 mm (5.25")292 mm (11.5")184 mm (7.25")07514-12 (L/S® Easy-Load® head)230 VAC; 50/60 Hz; 0.8 A600 rpmPainted steel and ABS plasticLog in Para disponibilidadEach (1UN)$44,410.00Supplier Number: 77910-75Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77910-75Model77910-75No. of channels1ChannelsNo. of rollers4Control typeAnalogHousing materialMin. flow rate per channel56 ml/minMax. flow rate per channel1700 ml/minDrive IP ratingIP 22Accuracy±5%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07555-05 (L/S® Analog Variable-Speed Console Drive)Duty cycleContinuousInputMin. RPM20 rpmOpen-head sensorNoPump seriesL/SRemote I/OStart/StopReversible motorYesTubing06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)Tubing sizes acceptedL/S® 15, 24Washdown capableNoCurrent0.8 AFrequency50/60 HzVoltage230 VACWarranty2 years2 year warranty - MFLX77715-90Masterflex L/S® Variable-speed pump system with easy-load thin-wall pump head and handheld remote, 600 rpm, 115 VAC (no open-head sensor), stainless steel rotor materialMasterflex L/S® Variable-speed pump system with easy-load thin-wall pump head and handheld remote, 600 rpm, 115 VAC (no open-head sensor), stainless steel rotor materialUS—————90 - 130 VAC, 50/60 Hz20 - 600—Log in Para disponibilidadEach (1UN)$47,780.00Supplier Number: 77715-90Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77715-90ModelNo. of channelsChannels1No. of rollers4; stainless steelControl typeHousing materialPolyarylamide (PARA)Min. flow rate per channel0.06 ml/minMax. flow rate per channel3.8 ml/minDrive IP ratingAccuracyAnti-dripCloud connectedDigital dispensingDriveDuty cycleInputStart/Stop (remote inputs)Min. RPMOpen-head sensorPump seriesL/SRemote I/OReversible motorTubingTubing sizes acceptedWashdown capableCurrentFrequencyVoltageWarranty2 years2 year warranty - MFLX77715-91Masterflex L/S® variable-speed pump system with easy-load thin-wall pump head and handheld remote, 600 rpm, 230 vac (no open-head sensor), stainless steel rotor materialMasterflex L/S® variable-speed pump system with easy-load thin-wall pump head and handheld remote, 600 rpm, 230 vac (no open-head sensor), stainless steel rotor materialUS—————230 VAC, 50/60 Hz20 - 600—Log in Para disponibilidadEach (1UN)$47,780.00Supplier Number: 77715-91Especificaciones:2 year warranty
More
Specifications
Cat. No.MFLX77715-91ModelNo. of channelsChannels1No. of rollers4; stainless steelControl typeHousing materialPolyarylamide (PARA)Min. flow rate per channel0.06 ml/minMax. flow rate per channel3.8 ml/minDrive IP ratingAccuracyAnti-dripCloud connectedDigital dispensingDriveDuty cycleInputStart/Stop (remote inputs)Min. RPMOpen-head sensorPump seriesL/SRemote I/OReversible motorTubingTubing sizes acceptedWashdown capableCurrentFrequencyVoltageWarranty2 years2 year warranty
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Footswitch, DB9 male, for use with drivesFootswitch, DB9 male, for use with drivesMFLX77595-35Log in Para disponibilidadEach (1UN) - $7,900.00
Información del producto
- Catalog #AvailabilityPrice
- Handheld Remote Controller; DB9(M) Connector; 6 ft (1,8 m) CableHandheld Remote Controller; DB9(M) Connector; 6 ft (1,8 m) CableMFLX07528-80Log in Para disponibilidadEach (1UN) - $9,600.00