Masterflex® I/P® Digital Precision Modular Drive Replacement Parts, Avantor®
Make the most of your Masterflex® I/P® Digital Precision Modular Drive with these replacement parts.
- Optimize performance of your Masterflex pumps
Keep your Masterflex® I/P® modular system running smoothly with these replacement parts.
- Environmentally Preferable:
- Modelo:
- 07594-14
- 07594-12
- 07594-02
- 07594-04
- Controller height:
- 119 mm (4.75")
- 140 mm (5.5")
- Controller length:
- 285 mm (10.2031")
- 263 mm (10.4062")
- Controller width:
- 279 mm (11")
- 211 mm (8.4062")
- Motor:
- 280 W
- 280 W
- Motor hp:
- ³/₈
- ³/₈
- Pump series:
- I/P
- I/P
- I/P
- I/P
- Speed control:
- ±0.1%
- ±0.1%
- Controller IP rating:
- IP 66
- IP 33
- Motor height:
- 119 mm (4.75")
- 165 mm (6.5")
- Motor IP rating:
- IP 66
- IP 33
- Motor length:
- 285 mm (10.1562")
- 362 mm (14.25")
- Motor width:
- 279 mm (11")
- 178 mm (7")
- Garantía:
- 2 years
- 2 years
- 2 years
- 2 years
Frequently Bought Together