Masterflex® L/S® Digital Modular Drives with Remote I/O Replacement Parts, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Make the most of your Masterflex drives replacement parts and other accessories.
- Extend the life of your Masterflex pump with these replacement parts
Choose replacement motors based on your Masterflex Modular System.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX07557-04
- MFLX07557-02
- MFLX07557-74
- MFLX07553-11
- MFLX07557-64
- MFLX07557-62
- MFLX07557-72
- MFLX07557-14
- MFLX07557-12
- MFLX07575-02
- Modelo:
- 07557-04
- 07557-02
- 07557-64
- 07557-62
- 07557-72
- 07557-14
- 07557-12
- Controller height:
- 140 mm (5.5")
- 119 mm (4.75")
- 119 mm (4.75")
- 140 mm (5.5")
- Controller length:
- 263 mm (10.4062")
- 285 mm (10.2031")
- 285 mm (10.2031")
- 263 mm (10.4062")
- Controller width:
- 211 mm (8.4062")
- 279 mm (11")
- 279 mm (11")
- 211 mm (8.4062")
- Motor:
- 75 W
- 75 W
- 75 W
- 75 W
- Motor hp:
- 1/10
- 1/10
- 1/10
- 1/10
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Speed control:
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- Washdown capable:
- No
- No
- Yes
- Yes
- Yes
- Yes
- No
- No
- Controller IP rating:
- IP 33
- IP 66
- IP 66
- IP 33
- Motor height:
- 114 mm (4.5")
- 152 mm (6")
- 152 mm (6")
- 114 mm (4.5")
- Motor IP rating:
- IP 33
- IP 66
- IP 66
- IP 33
- Motor length:
- 273 mm (10.75")
- 395 mm (15.8125")
- 395 mm (15.8125")
- 273 mm (10.75")
- Motor width:
- 111 mm (4.375")
- 178 mm (7")
- 178 mm (7")
- 111 mm (4.375")
- Garantía:
- 2 years
- 2 years
- 2 years
- 1 year
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years