Masterflex® L/S® High-Performance Pump Head Rotor Assembly Clip, Avantor®
L/S High-Performance Rotor Assembly Clip for 77250-62 /-82
Catalog # MFLX77250-69
Supplier: Avantor Fluid Handling
: 77250-69
Masterflex® L/S® High-Performance Pump Head Rotor Assembly Clip, Avantor®
Catalog # MFLX77250-69
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Get the most out of your Masterflex® L/S® High-Performance Pump Head with these replacement parts.
- Suitable for use with High-Performance pump heads 77250-62 and -82
Replacement parts to keep your pump head operating even longer.
Especificaciones
- Description:L/S High-Performance Rotor Assembly Clip for 77250-62 /-82
- Warranty:2 years
- Cat. No.:MFLX77250-69
- Pump series:L/S