ContactUsLinkComponent
Masterflex® Drum Pump ODP Motors, Avantor®
Masterflex® Drum Pump Replacement Motor, ODP, 32GPM; 115V
Catalog # MFLX70604-00
Proveedor:  VWR International
undefined
Masterflex® Drum Pump Replacement Motor, ODP, 32GPM; 115V
Catalog # MFLX70604-00
Proveedor:  VWR International
Especificaciones
  • Tasa de flujo máxima:
    121.13 L/min (32 gal./min)
  • Potencia:
    115 VAC; 60 Hz
  • Viscosidad máx.:
    750 cP
  • Descripción:
    Drum Pump ODP Motor, 115 VAC
  • Motor hp:
    1-1/10
  • Motor type:
    ODP
  • Garantía:
    4 months
  • Referencia:
    MFLX70604-00
  • Max. Motor RPM:
    10000 rpm
Especificaciones
Acerca de este artículo
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
  • Flow rate at 32 GPM
This drum pump motor is designed to transfer a variety of materials from 55 gallon drums and tanks. Drum pump tubes are sold separatley, making this drum pump customizable to your specific application. Pump tubes are available in multiple materials and lengths, model numbers 70604-42, -44, -52, -54, -62, -64, -66, and -68. Barrel adapter 70604-92 sold separately.
The motor has thermal overload protection which stops the pump in case of overload, allowing it to cool down.
  • Tasa de flujo máxima:
    121.13 L/min (32 gal./min)
  • Potencia:
    115 VAC; 60 Hz
  • Viscosidad máx.:
    750 cP
  • Descripción:
    Drum Pump ODP Motor, 115 VAC
  • Environmentally Preferable:
  • Motor hp:
    1-1/10
  • Motor type:
    ODP
  • Garantía:
    4 months
  • Referencia:
    MFLX70604-00
  • Max. Motor RPM:
    10000 rpm