ContactUsLinkComponent
Masterflex® Luer Fittings, High-Pressure Stopcock, Avantor®
Masterflex® Luer Fittings, High-Pressure Stopcock, Avantor®
Supplier:  VWR International
Certificates
undefined
Masterflex® Luer Fittings, High-Pressure Stopcock, Avantor®
Supplier:  VWR International
Specifications
  • Presión máx.:
  • 1050 psi
  • Wetted materials:
  • Polycarbonate
  • Garantía:
  • Out of box