Masterflex® Particle Tolerant Solenoid Valves, Avantor®
Supplier: CLARK SOLUTIONS
Certificates
About this item
Particles will not cause damage.
- Special elastomer diaphragm and seals are forgiving of particulate matter that would typically damage a plastic valve seat
- Inert materials — excellent chemical resistance
- Molded PEEK valve bodies
- Excellent choice for demanding low-flow applications where minimum size and internal volume are important
These Masterflex® particle tolerant solenoid valves feature a apecial Perfluor™ soft elastomer diaphragm and seals, which are forgiving of particulate matter that would typically damage a plastic valve seat. Valves are constructed with a PEEK body, PTFE diaphragm, Perfluor seat and seal, and ceramic stem. Choose from two-way normally closed valves, two-way normally open valves, and three-way valve options.
- Environmentally Preferable:
Specifications
- Modelo:
- 98305-24
- 98305-42
- 98305-34
- 98305-26
- 98305-32
- 98305-36
- Presión máx.:
- Common: 29 psi; NC/NO: 14 psi
- Common: 29 psi; NC/NO: 7 psi
- Inlet: 29 psi; Outlet: 14 psi
- Common: 29 psi; NC/NO: 7 psi
- Max. temperature:
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- Orifice:
- 2 mm (0.0787")
- 2 mm (0.0787")
- 1.2 mm (0.0472")
- 2 mm (0.0787")
- 2 mm (0.0787")
- 1.2 mm (0.0472")
- Alimentación:
- 12 VDC, 2.6 W
- 24 VDC, 2.6 W
- 24 VDC, 2.5 W
- 24 VDC, 2.6 W
- 12 VDC, 2.5 W
- 12 VDC, 2.5 W
- Tiempo de respuesta:
- 15 ms
- 15 ms
- 15 ms
- 15 ms
- 15 ms
- 15 ms
- Wetted materials:
- PEEK body, PTFE diaphragm, Perfluor® seat/seal; ceramic stem
- PEEK body, PTFE diaphragm, Perfluor® seat/seal; ceramic stem
- PEEK body, PTFE diaphragm, Perfluor® seat/seal; ceramic stem
- PEEK body, PTFE diaphragm, Perfluor® seat/seal; ceramic stem
- PEEK body, PTFE diaphragm, Perfluor® seat/seal; ceramic stem
- PEEK body, PTFE diaphragm, Perfluor® seat/seal; ceramic stem
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year