Orders LinkContactUsLinkComponent
Masterflex® Check Valves, PTFE, Avantor®
Masterflex® Check Valves, PTFE, Avantor®
Supplier:   VWR International
Masterflex® Check Valves, PTFE, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Check Valves, PTFE, Avantor®
1 Options Available Below
Product Details & Documents
Use with caustic and high-purity liquids in many applications.
  • Constructed of durable PTFE
Designed for use with corrosive fluids of varying viscosities in chemical, microelectronics, and petroleum industries.
Specifications for Product Family
Especificaciones
  • Model:
  • 98515-14
  • 98515-16
  • 98515-18
  • Max. pressure:
  • 50 psi
  • 50 psi
  • 50 psi
  • Max. temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Orifice:
  • 6.35 mm (0.25")
  • 9.525 mm (0.375")
  • 12.7 mm (0.5")
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Connection Type
    Inlet Port
    Outlet Port
    Port Size
    Availability
    Price
  • MFLX98515-14
    PTFE Check valve, ¹/₄" Orifice, ¹/₄" Female NPT
    PTFE Check valve, ¹/₄" Orifice, ¹/₄" Female NPT
    NPT(F)
    ¼" NPT(F)
    ¼" NPT(F)
    0.25 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $10,840.00
    $10,840.00
    Each (1UN)
       $10,840.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX98515-14
    Model
    98515-14
    Max. pressure
    50 psi
    Max. temperature
    100 °C (212 °F)
    Orifice
    6.35 mm (0.25")
    Warranty
    1 year
  • MFLX98515-16
    PTFE Check valve, ³/₈" Orifice, ³/₈" Female NPT
    PTFE Check valve, ³/₈" Orifice, ³/₈" Female NPT
    NPT(F)
    ⅜" NPT(F)
    ⅜" NPT(F)
    0.375 NPT(F)
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX98515-16
    Model
    98515-16
    Max. pressure
    50 psi
    Max. temperature
    100 °C (212 °F)
    Orifice
    9.525 mm (0.375")
    Warranty
    1 year
  • MFLX98515-18
    PTFE Check valve, ¹/₂" Orifice, ¹/₂" Female NPT
    PTFE Check valve, ¹/₂" Orifice, ¹/₂" Female NPT
    NPT(F)
    ½" NPT(F)
    ½" NPT(F)
    0.5 NPT(F)
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX98515-18
    Model
    98515-18
    Max. pressure
    50 psi
    Max. temperature
    100 °C (212 °F)
    Orifice
    12.7 mm (0.5")
    Warranty