Masterflex® Transfer Tubing Spool, Polytetrafluoroethylene (PTFE), Avantor®
Proveedor: VWR International
Certificados
Acerca de este artículo
Available in spooled rolls for increased versatility.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.
- Environmentally Preferable:
Especificaciones
- Modelo:
- 95231-14
- 95231-02
- 95231-06
- 95231-12
- 95231-10
- 95231-08
- 95231-00
- Radio de doblez:
- 50.8 mm (2")
- 12.7 mm (0.5")
- 25.4 mm (1")
- 63.5 mm (2.5")
- 25.4 mm (1")
- 44.5 mm (1.75")
- 12.7 mm (0.5")
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 30.5 m (100 ft.)
- 76.2 m (250 ft.)
- 76.2 m (250 ft.)
- 76.2 m (250 ft.)
- 76.2 m (250 ft.)
- 76.2 m (250 ft.)
- 76.2 m (250 ft.)
- Presión máx.:
- 5.4 bar (79 psi)
- 23.4 bar (339 psi)
- 9.9 bar (144 psi)
- 6.3 bar (92 psi)
- 14.1 bar (204 psi)
- 7.9 bar (114 psi)
- 23.2 bar (336 psi)
- Max. temperature:
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- 287 °C (550 °F)
- Min. temperature:
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- –267 °C (–450 °F)
- Regulatory approvals:
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 5.4 bar (79 psi)
- 23.4 bar (339 psi)
- 9.9 bar (144 psi)
- 6.3 bar (92 psi)
- 14.1 bar (204 psi)
- 7.9 bar (114 psi)
- 23.2 bar (336 psi)
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year