Masterflex® Transfer Tubing, Ester-Based Polyurethane, Avantor®
Worry-free petroleum-based transfer.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
The tubes are ideal for high-purity, general laboratory, petroleum-based transfer. Characteristics include plasticizer-free, low extractables, good at low temperatures, good resistance to petroleum-based chemicals, and high tensile strength. Not recommended for use with aqueous solutions. Temperature range is -70 to 185 °F (-56 to 85 °C). Can be sterilized with ethylene oxide.
Worry-free petroleum-based transfer.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
The tubes are ideal for high-purity, general laboratory, petroleum-based transfer. Characteristics include plasticizer-free, low extractables, good at low temperatures, good resistance to petroleum-based chemicals, and high tensile strength. Not recommended for use with aqueous solutions. Temperature range is -70 to 185 °F (-56 to 85 °C). Can be sterilized with ethylene oxide.
- Catalog No:
- MFLX95680-36
- MFLX95680-32
- MFLX95680-14
- MFLX95680-12
- MFLX95680-16
- MFLX95680-04
- MFLX95680-02
- MFLX95680-00
- MFLX95680-50
- MFLX95680-46
- MFLX95680-22
- MFLX95680-26
- MFLX95680-06
- MFLX95680-08
- MFLX95680-42
- MFLX95680-20
- MFLX95680-28
- Model:
- 95680-36
- 95680-32
- 95680-14
- 95680-12
- 95680-16
- 95680-04
- 95680-02
- 95680-00
- 95680-50
- 95680-46
- 95680-22
- 95680-26
- 95680-06
- 95680-08
- 95680-42
- 95680-20
- 95680-28
- Bend radius:
- 44.45 mm (1.75")
- 73.025 mm (2.875")
- 23.8125 mm (0.9375")
- 39.6875 mm (1.5625")
- 17.4625 mm (0.6875")
- 7.9375 mm (0.3125")
- 12.7 mm (0.5")
- 4.7625 mm (0.1875")
- 130.175 mm (5.125")
- 82.55 mm (3.25")
- 44.45 mm (1.75")
- 26.9875 mm (1.0625")
- 25.4 mm (1")
- 15.875 mm (0.625")
- 60.325 mm (2.375")
- 49.2125 mm (1.9375")
- 42.8625 mm (1.6875")
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- 30.5 m (100 ft.)
- Max. pressure:
- 4.1 bar (60 psi)
- 2.5 bar (36 psi)
- 4.6 bar (67 psi)
- 2.3 bar (33 psi)
- 5.1 bar (74 psi)
- 6.9 bar (100 psi)
- 5 bar (72 psi)
- 6.9 bar (100 psi)
- 2.8 bar (40 psi)
- 3.5 bar (51 psi)
- 4.1 bar (59 psi)
- 6.9 bar (100 psi)
- 1.9 bar (27 psi)
- 5.1 bar (74 psi)
- 4.1 bar (60 psi)
- 4.5 bar (65 psi)
- 5.1 bar (74 psi)
- Max. temperature:
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- Max. vacuum:
- 29,9" Hg
- 20" Hg
- 29,9" Hg
- 20" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 15" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- 29,9" Hg
- Min. temperature:
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 4.1 bar (60 psi)
- 2.5 bar (36 psi)
- 4.6 bar (67 psi)
- 2.3 bar (33 psi)
- 5.1 bar (74 psi)
- 6.9 bar (100 psi)
- 5 bar (72 psi)
- 6.9 bar (100 psi)
- 2.8 bar (40 psi)
- 3.5 bar (51 psi)
- 4.1 bar (59 psi)
- 6.9 bar (100 psi)
- 1.9 bar (27 psi)
- 5.1 bar (74 psi)
- 4.1 bar (60 psi)
- 4.5 bar (65 psi)
- 5.1 bar (74 psi)
Product Family Options
- DescriptionTubing int.ØWall thicknessTubing ext.ØTubing MaterialLengthColor
- Transfer Tubing, Ester-Based Polyurethane, ¹/₂" ID x ³/₄" OD; 100 Ft12.8 mm (0.5")3.2 mm (0.125")19.2 mm (0.75")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$16,010.00
Specifications
Cat. No.MFLX95680-36Model95680-36Bend radius44.45 mm (1.75")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure4.1 bar (60 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4.1 bar (60 psi) - Transfer Tubing, Ester-Based Polyurethane, ¹/₂" ID x 5/8" OD; 100 Ft12.8 mm (0.5")1.6 mm (0.0625")16 mm (0.625")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$8,210.00
Specifications
Cat. No.MFLX95680-32Model95680-32Bend radius73.025 mm (2.875")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure2.5 bar (36 psi)Max. temperature93 °C (200 °F)Max. vacuum20" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure2.5 bar (36 psi) - Transfer Tubing, Ester-Based Polyurethane, ¹/₄" ID x ³/₈" OD; 100 Ft6.4 mm (0.25")1.6 mm (0.0625")9.6 mm (0.375")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$5,110.00
Specifications
Cat. No.MFLX95680-14Model95680-14Bend radius23.8125 mm (0.9375")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure4.6 bar (67 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4.6 bar (67 psi) - Transfer Tubing, Ester-Based Polyurethane, ¹/₄" ID x 5/16" OD; 100 Ft6.4 mm (0.25")0.8 mm (0.313")8 mm (0.3125")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$4,370.00
Specifications
Cat. No.MFLX95680-12Model95680-12Bend radius39.6875 mm (1.5625")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure2.3 bar (33 psi)Max. temperature93 °C (200 °F)Max. vacuum20" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure2.3 bar (33 psi) - Transfer Tubing, Ester-Based Polyurethane, ¹/₄" ID x 7/16" OD; 100 Ft6.4 mm (0.25")2.4 mm (0.0937")11.2 mm (0.4375")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$9,170.00
Specifications
Cat. No.MFLX95680-16Model95680-16Bend radius17.4625 mm (0.6875")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure5.1 bar (74 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5.1 bar (74 psi) - Transfer Tubing, Ester-Based Polyurethane, ¹/₈" ID x ¹/₄" OD; 100 Ft3.2 mm (0.125")1.6 mm (0.0625")6.4 mm (0.25")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$3,570.00
Specifications
Cat. No.MFLX95680-04Model95680-04Bend radius7.9375 mm (0.3125")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure6.9 bar (100 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure6.9 bar (100 psi) - Transfer Tubing, Ester-Based Polyurethane, ¹/₈" ID x ³/₁₆" OD; 100 Ft3.2 mm (0.125")0.8 mm (0.313")4.8 mm (0.1875")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$1,880.00
Specifications
Cat. No.MFLX95680-02Model95680-02Bend radius12.7 mm (0.5")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure5 bar (72 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5 bar (72 psi) - Transfer Tubing, Ester-Based Polyurethane, 1/16" ID x ¹/₈" OD; 100 Ft1.6 mm (0.0625")0.8 mm (0.313")3.2 mm (0.125")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$1,540.00
Specifications
Cat. No.MFLX95680-00Model95680-00Bend radius4.7625 mm (0.1875")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure6.9 bar (100 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure6.9 bar (100 psi) - Transfer Tubing, Ester-Based Polyurethane, 1" ID x 1-¹/₄" OD; 100 Ft25.4 mm (1")3.2 mm (0.125")31.8 mm (1.25")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$28,470.00
Specifications
Cat. No.MFLX95680-50Model95680-50Bend radius130.175 mm (5.125")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure2.8 bar (40 psi)Max. temperature93 °C (200 °F)Max. vacuum15" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure2.8 bar (40 psi) - Transfer Tubing, Ester-Based Polyurethane, ³/₄" ID x 1" OD; 100 Ft19.2 mm (0.75")3.2 mm (0.125")25.4 mm (1")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$19,040.00
Specifications
Cat. No.MFLX95680-46Model95680-46Bend radius82.55 mm (3.25")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure3.5 bar (51 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure3.5 bar (51 psi) - Transfer Tubing, Ester-Based Polyurethane, ³/₈" ID x ¹/₂" OD; 100 Ft9.6 mm (0.375")1.6 mm (0.0625")12.8 mm (0.5")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$6,420.00
Specifications
Cat. No.MFLX95680-22Model95680-22Bend radius44.45 mm (1.75")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure4.1 bar (59 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4.1 bar (59 psi) - Transfer Tubing, Ester-Based Polyurethane, ³/₈" ID x 5/8" OD; 100 Ft9.6 mm (0.375")3.2 mm (0.125")16 mm (0.625")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$12,230.00
Specifications
Cat. No.MFLX95680-26Model95680-26Bend radius26.9875 mm (1.0625")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure6.9 bar (100 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure6.9 bar (100 psi) - Transfer Tubing, Ester-Based Polyurethane, ³/₁₆" ID x ¹/₄" OD; 100 Ft4.8 mm (0.1875")0.8 mm (0.313")6.4 mm (0.25")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$2,400.00
Specifications
Cat. No.MFLX95680-06Model95680-06Bend radius25.4 mm (1")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure1.9 bar (27 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure1.9 bar (27 psi) - Transfer Tubing, Ester-Based Polyurethane, ³/₁₆" ID x 5/16" OD; 100 Ft4.8 mm (0.1875")1.6 mm (0.0625")8 mm (0.3125")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$4,290.00
Specifications
Cat. No.MFLX95680-08Model95680-08Bend radius15.875 mm (0.625")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure5.1 bar (74 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5.1 bar (74 psi) - Transfer Tubing, Ester-Based Polyurethane, 5/8" ID x 7/8" OD; 100 Ft16 mm (0.625")3.2 mm (0.125")22.4 mm (0.875")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$17,400.00
Specifications
Cat. No.MFLX95680-42Model95680-42Bend radius60.325 mm (2.375")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure4.1 bar (60 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4.1 bar (60 psi) - Transfer Tubing, Ester-Based Polyurethane, 5/16" ID x 7/16" OD; 100 Ft8 mm (0.3125")1.6 mm (0.0625")1.2 mm (0.4375")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$7,530.00
Specifications
Cat. No.MFLX95680-20Model95680-20Bend radius49.2125 mm (1.9375")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure4.5 bar (65 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure4.5 bar (65 psi) - Transfer Tubing, Ester-Based Polyurethane, 7/16" ID x 5/8" OD; 100 Ft11.2 mm (0.4375")2.4 mm (0.0937")16 mm (0.625")Ester-based PU30.5 m (100 ft.)NaturalEach (100Pie)$11,330.00
Specifications
Cat. No.MFLX95680-28Model95680-28Bend radius42.8625 mm (1.6875")ClarityTranslucentLength per pack30.5 m (100 ft.)Max. pressure5.1 bar (74 psi)Max. temperature93 °C (200 °F)Max. vacuum29,9" HgMin. temperature–73 °C (–100 °F)Sterilisation method autoclaveNoSterilisation method EtOYesSterilisation method gammaNoWorking pressure5.1 bar (74 psi)