Masterflex® Low-Flow Peristaltic Pump Replacement Tubing Sets, Avantor®
Supplier: VWR International
Certificates
About this item
Choose tubing based on chemical compatibility, flow rate, and your current pump.
- Global market leader in range of formulations for most extensive chemical compatibility
Choose from silicone, norprene and viton replacement tubing for your peristaltic pump.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX73160-56
- MFLX73160-58
- MFLX73160-54
- Modelo:
- 73160-56
- 73160-58
- 73160-54
- Inlet connection:
- 0.8. 1.6. 3.0 mm ID tubing
- 0.8. 1.6. 3.0 mm ID tubing
- 0.8. 1.6. 3.0 mm ID tubing
- Presión máx.:
- 2.06 bar (30 psi)
- 0.7 bar (10 psi)
- 0.7 bar (10 psi)
- Max. temperature:
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- Velocidad mín. de flujo:
- 0.03 ml/min
- 0.03 ml/min
- 0.03 ml/min
- Outlet connection:
- 0.8, 1.6, 3.0 mm int.Ø tubing
- 0.8, 1.6, 3.0 mm int.Ø tubing
- 0.8, 1.6, 3.0 mm int.Ø tubing
- Wetted materials:
- Polypropylene, Norprene
- Polypropylene, Viton®
- Polypropylene, silicone
- Garantía:
- 3 Months
- 3 Months
- 3 Months