Masterflex® L/S® Tube Sets, PTFE, Avantor®
Proveedor: Avantor Fluid Handling
Certificados
Acerca de este artículo
Excellent chemical resistance and chemically inert.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
For use with Masterflex® L/S® PTFE Tubing Pump Head
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX77390-60
- MFLX77390-50
- Length per pack:
- 0.38 m (1.25 ft.)
- 0.38 m (1.25 ft.)
- Presión máx.:
- 6.9 bar (100 psi)
- 6.9 bar (100 psi)
- Min. temperature:
- –240 °C (–400 °F)
- –240 °C (–400 °F)
- Max. temperature:
- 260 °C (500 °F)
- 260 °C (500 °F)
- Min. temperature (dynamic):
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- Max. temperature (dynamic):
- 150 °C (302 °F)
- 150 °C (302 °F)
- Pump series:
- L/S
- L/S
- Regulatory approvals:
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- Sterilisation:
- Ethylene Oxide
- Ethylene Oxide
- Sterilisation method autoclave:
- No
- No
- Sterilisation method EtO:
- Yes
- Yes
- Sterilisation method gamma:
- No
- No
Especificaciones
Información relacionada
Frequently Bought Together