Orders LinkContactUsLinkComponent
Aalborg Digital Flowmeter Cables and Adapters
Aalborg Digital Flowmeter Cables and Adapters
Aalborg Digital Flowmeter Cables and Adapters
Certificates
Aalborg Digital Flowmeter Cables and Adapters
15 Options Available Below
Product Details & Documents
Displays instantaneous, total and accumulated flow rates.
  • Built-in Flow Linearizer (10 point linearization of the flow curve)
  • Up to 47 different volumetric and mass flow engineering units
  • Large 13mm (0.51") digits for flow rate and 5.5mm (0.21") for Total
  • Digital RS-232 interface (multi-drop capability of up to 64 devices requires RS-485)
  • Compact design for unit mount, panel mount, wall mount or field mount applications
  • User-programmable, optically-isolated pulse output
  • Two programmable, optically-isolated, digital outputs
  • Flow controllers, set point command control via local LCD or digital interface
  • Programmable set point table with ramping up/down capability up to 16 steps
  • Free configuration and monitoring utility software
Use these totalizers for flowmeters and controllers with a voltage or current input and output interface where flow indications / control and totalizers, or alarm functions are required. These flow totalizers can be used when re-transmission of the flow rate and/or totalizer functions via optically-isolated pulse output or serial communication is desired.Local or programmable set point control for flow controllers are easily done via keypad. Activation of user-supplied equipment via programmable optically-isolated digital outputs are easily done when flow alarms or totalizers events are active.
The graphical LCD display has large ½” (13 mm) digits for flow rate and 0.21" (5.5 mm) for total and can be set by you to simultaneously show different combination of the flow parameters: flow rate, totalizers, flow alarms, and diagnostic events. All configuration parameter settings are easily accessed via a simple user-interface menu driven by a 6 button key-pad which can be password-protected.
Totalizer kits, below, enable you to power your totalizer depending on your input/output power requirements, for both flowmeters and controllers, and are required if you have no power or communication cables. Kits 32650-92 and -94 are mounting brackets only, and require user-supplied custom cables.
Specifications for Product Family
Especificaciones
  • Model:
  • 03277-00
  • 03277-05
  • 32650-22
  • 32650-60
  • 32650-84
  • 32650-86
  • 32662-50
  • 32662-55
  • 32662-60
  • 32662-65
  • 32662-70
  • 32755-90
  • 33401-90
  • 33401-91
  • 32650-65
  • 32650-80
  • 32650-88
  • 32650-92
  • 32650-94
  • 32650-96
  • 33401-92
  • 32650-90
  • Connection:
  • 9-pin D-Sub connector
  • 12-pin M16 shielded cable
  • Communication interface:
  • RS-232
  • Display type:
  • 128 × 64 graphic LCD with 6 tactical push buttons
  • Min. flow rate:
  • 0 L/min
  • 0 L/min
  • 0 L/min
  • Min. operating temperature:
  • –10 °C (15 °F)
  • Max. operating temperature:
  • 70 °C (158 °F)
  • Input:
  • Jumper selectable 0-5 VDC, 5-10 VDC, or 4-20 mA
  • Output:
  • Voltage, 0 to 5 VDC or 4 to 20 mA
  • Warranty:
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Electrical
    Length
    Availability
    Price
  • MFLX03277-00
    Mass flowmeter power supply/adapter
    Mass flowmeter power supply/adapter
    110 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,070.00
    $1,070.00
    Each (1UN)
       $1,070.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX03277-00
    Model
    03277-00
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX03277-05
    Mass flowmeter power supply/adapter
    Mass flowmeter power supply/adapter
    220 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,500.00
    $1,500.00
    Each (1UN)
       $1,500.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX03277-05
    Model
    03277-05
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-22
    Totalizer for gas mass flowmeters and controllers with input and output
    Totalizer for gas mass flowmeters and controllers with input and output
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $11,900.00
    $11,900.00
    Each (1UN)
       $11,900.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-22
    Model
    32650-22
    Connection
    9-pin D-Sub connector
    Communication interface
    RS-232
    Display type
    128 × 64 graphic LCD with 6 tactical push buttons
    Min. flow rate
    Min. operating temperature
    –10 °C (15 °F)
    Max. operating temperature
    70 °C (158 °F)
    Input
    Jumper selectable 0-5 VDC, 5-10 VDC, or 4-20 mA
    Output
    Voltage, 0 to 5 VDC or 4 to 20 mA
    Warranty
  • MFLX32650-60
    Output cable for 4 to 20 mA signal
    Output cable for 4 to 20 mA signal
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $654.00
    $654.00
    Each (1UN)
       $654.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-60
    Model
    32650-60
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-84
    Flow meter mounting kit, flat wire cable between GFM and TIO9 pin D-connectors
    Flow meter mounting kit, flat wire cable between GFM and TIO9 pin D-connectors
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $492.00
    $492.00
    Each (1UN)
       $492.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-84
    Model
    32650-84
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-86
    GFC flowmeter mounting kit with communication branch
    GFC flowmeter mounting kit with communication branch
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,760.00
    $1,760.00
    Each (1UN)
       $1,760.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-86
    Model
    32650-86
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32662-50
    Flowmeter power supply/adapter, GFC series
    Flowmeter power supply/adapter, GFC series
    110 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,430.00
    $1,430.00
    Each (1UN)
       $1,430.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32662-50
    Model
    32662-50
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32662-55
    Flowmeter power supply/adapter, GFC series Euro
    Flowmeter power supply/adapter, GFC series Euro
    220 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,650.00
    $1,650.00
    Each (1UN)
       $1,650.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32662-55
    Model
    32662-55
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32662-60
    Flowmeter power supply/adapter, GFC series UK
    Flowmeter power supply/adapter, GFC series UK
    220 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,130.00
    $2,130.00
    Each (1UN)
       $2,130.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32662-60
    Model
    32662-60
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32662-65
    Cable with DB15(F) connectors with bare wire ends
    Cable with DB15(F) connectors with bare wire ends
    3 ft
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (3Pie)
    Each (3Pie) - $1,110.00
    $1,110.00
    Each (3Pie)
       $1,110.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32662-65
    Model
    32662-65
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32662-70
    Extension cable for remote reading
    Extension cable for remote reading
    3 ft
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (3Pie)
    Each (3Pie) - $492.00
    $492.00
    Each (3Pie)
       $492.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32662-70
    Model
    32662-70
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32755-90
    Power cable, 12 pin shielded M16
    Power cable, 12 pin shielded M16
    6 ft.
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (6Pie)
    Each (6Pie) - $3,890.00
    $3,890.00
    Each (6Pie)
       $3,890.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32755-90
    Model
    32755-90
    Connection
    12-pin M16 shielded cable
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
    1 year
  • MFLX33401-90
    Power and communication cable with 15-pin D-connector
    Power and communication cable with 15-pin D-connector
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,090.00
    $1,090.00
    Each (1UN)
       $1,090.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX33401-90
    Model
    33401-90
    Connection
    Communication interface
    Display type
    Min. flow rate
    0 L/min
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX33401-91
    Power and communication cable with 15-pin D-connector
    Power and communication cable with 15-pin D-connector
    115 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,430.00
    $1,430.00
    Each (1UN)
       $1,430.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX33401-91
    Model
    33401-91
    Connection
    Communication interface
    Display type
    Min. flow rate
    0 L/min
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-65
    Output cable for 0 to 5 VDC signal
    Output cable for 0 to 5 VDC signal
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $654.00
    $654.00
    Each (1UN)
       $654.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-65
    Model
    32650-65
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-80
    GFM flowmeter mounting kit with communication branch
    GFM flowmeter mounting kit with communication branch
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-80
    Model
    32650-80
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-88
    GFC flowmeter mounting kit with communication branch
    GFC flowmeter mounting kit with communication branch
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-88
    Model
    32650-88
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-92
    Flow meter and totalizer mounting kit, no cables
    Flow meter and totalizer mounting kit, no cables
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-92
    Model
    32650-92
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-94
    Flow controller and totalizer mounting kit, no cables
    Flow controller and totalizer mounting kit, no cables
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-94
    Model
    32650-94
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-96
    Flow totalizer accessory, 9 conductor round shielded cable
    Flow totalizer accessory, 9 conductor round shielded cable
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-96
    Model
    32650-96
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX33401-92
    Power supply with communication and 9-wire analog output
    Power supply with communication and 9-wire analog output
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX33401-92
    Model
    33401-92
    Connection
    Communication interface
    Display type
    Min. flow rate
    0 L/min
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
  • MFLX32650-90
    GFC flowmeter mounting kit
    GFC flowmeter mounting kit
    110 VAC, 24 VDC
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32650-90
    Model
    32650-90
    Connection
    Communication interface
    Display type
    Min. flow rate
    Min. operating temperature
    Max. operating temperature
    Input
    Output
    Warranty
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • GFC flowmeter mounting kit with communication branch
    GFC flowmeter mounting kit with communication branch
    MFLX32650-86
    Log in Para disponibilidad
    Each (1UN) - $1,760.00
Información del producto
  • Catalog #
    Availability
    Price
  • Flow meter mounting kit, flat wire cable between GFM and TIO9 pin D-connectors
    Flow meter mounting kit, flat wire cable between GFM and TIO9 pin D-connectors
    MFLX32650-84
    Log in Para disponibilidad
    Each (1UN) - $492.00