ContactUsLinkComponent
Masterflex® Union Fittings, Hose Barb, Straight, PVDF, Avantor®
undefined
Masterflex® Union Fittings, Hose Barb, Straight, PVDF, Avantor®
  • Orifice dimensions:
  • 14.29 mm (0.5625")
  • 11.9 mm (0.46875")
  • 0.09375"
  • 0.7925 mm (0.03125")
  • 0.0625"
  • 3.175 mm (0.125")
  • 4.763 mm (0.1875")
  • 0.03125"
  • 6.35 mm (0.25")
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
Frequently Bought Together