Masterflex® Press-In Plug Fittings, Hose Barb, Straight, Avantor®
Proveedor: VWR International
Certificados
Acerca de este artículo
Protect against contaminants in tubing lines.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Use these barbed press-in-plugs to keep open tubing lines or tubing assemblies free of foreign contaminants and ensure process purity. Plugs are manufactured from 100% virgin materials in a ISO Class 8 (formerly Class 100,000) environment and include full material traceability. Available in PVDF or nylon. All plugs are sterilizable by gamma irradiation; nylon plugs may also be autoclaved.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX40615-45
- MFLX40615-44
- MFLX51519-46
- MFLX41519-35
- MFLX31220-11
- MFLX41519-43
- MFLX51519-38
- MFLX40615-43
- MFLX41519-44
- MFLX41519-36
- MFLX40615-38
- MFLX40615-40
- MFLX40615-39
- MFLX40615-42
- MFLX40615-41
- Modelo:
- 51519-46
- 41519-35
- 31220-11
- 41519-43
- 51519-38
- 41519-44
- 41519-36
- Sterilisation:
- Autoclave or gamma
- Garantía:
- 3 Months