Masterflex® Luer Lock Fittings, Stopcock, Avantor®
Build leakproof fluid handling systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
- Environmentally Preferable:
- Modelo:
- 31200-92
- 31200-91
- 31200-90
- Wetted materials:
- Black PVDF
- Black PVDF
- Natural PVDF, Polypropylene
- Black PVDF
Frequently Bought Together