Masterflex® Adapter Fittings, Sanitary Clamp to Female Threaded, Straight, Avantor®
Maintain sanitary conditions.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
- All fittings are compatible with Tri-Clamp® connections
- Excellent chemical resistance; low leachable characteristics
These fittings are ideal for use with the Masterflex L/S tubing pumps.
- Environmentally Preferable:
Frequently Bought Together