Orders LinkContactUsLinkComponent
Dissolved Oxygen, CHEMets Visual Kit, CHEMetrics
Dissolved Oxygen, CHEMets Visual Kit, CHEMetrics
Oxygen, dissolved CHEMets Refill, Rhodazine D, 0 - 40 ppb
Catalog # CXR-7540
Supplier:   CHEMetrics
Supplier #:  R-7540
Dissolved Oxygen, CHEMets Visual Kit, CHEMetrics
Catalog # CXR-7540
Supplier:  CHEMetrics
CAS Number:  
Dissolved Oxygen, CHEMets Visual Kit, CHEMetrics
Dissolved Oxygen, CHEMets Visual Kit, CHEMetrics
Oxygen, dissolved CHEMets Refill, Rhodazine D, 0 - 40 ppb
CAS Number:  
Price
$1,731.85
Log in for availability
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for low- to medium-range colorimetric analysis of water quality
CHEMets® test kits utilize self-filling ampules and vacuum pulls the sample up from a snap cup into the ampule when the tip is broken off. After tilting the ampule several times to mix the sample and reagent and waiting for the color to develop (usually 2 minutes or less), the analyst quantifies the result using the provided cylindrical comparator (for concentrations ≤1ppm) or flat comparator (for concentrations ≥1ppm).
The level of dissolved oxygen in natural waters is often a direct indication of quality, since aquatic plants produce oxygen, while microorganisms generally consume it as they feed on pollutants.
At low temperatures the solubility of oxygen is increased; during summer, saturation levels can be as low as 4 ppm. Dissolved oxygen (D.O.) is essential for the support of fish and other aquatic life and aids in the natural decomposition of organic matter. Waste treatment plants that employ aerobic digestion must maintain a level of at least 2 ppm dissolved oxygen.
Caution: WARNING! The product employing the Rhodazine D method can expose you to chemicals including methanol, which is known to the State of California to cause birth defects or other reproductive harm.
Specifications
Specifications
  • Chemical method:
    Rhodazine D
  • Description:
    Oxygen, dissolved CHEMets Refill
  • Range:
    0 - 40 ppb
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 