Measuring Cylinder Sets, Polypropylene, Round Base, United Scientific
470164-042
- Descripción:Measuring cylinder sets, PP, round base
- Referencia:470164-042
Durable set of graduated cylinder.
- Polypropylene
- Autoclavable
A convenient set of four graduated polypropylene cylinders in sizes 10 ml, 25 ml, 50 ml, and 100 ml.