Argos Technologies® Reusable Inoculating Loops without Handles, Antylia Scientific
Catalog # 76534-534
Supplier: ANTYLIA SCIENTIFIC
Specifications
- Esterilidad:Non sterile
- Volumen:1 µl
- Descripción:Calibrated inoculating loops
- Referencia:76534-534
- Diámetro interno:1.45 mm
- Calibrador:26
Specifications
About this item
Nichrome wire composition cools quickly and is less expensive than platinum.
- Select, inoculate, or transfer microbiological samples
Argos brand Inoculating Loops are made from high quality twisted nichrome wire. Loops have been designed to slide along agar surfaces without cutting or tearing.
Argos Inoculating Loops cool rapidly and are less expensive than platinum. Loops are supplied without handles and are available calibrated or non-calibrated.
Ordering information: Brass loop handles are sold separately as models 04396-40 and -41.