Membrane Filters with Grid, Sartorius
76500-654
- Ø:47 mm
- Tamaño de poro:0.45 µm
- Esterilidad:Sterile
- Tipo:131
- Max. temperature:130 °C
- Descripción:White with black grid, cellulose nitrate, 3 mm hydrophobic edge
- Grosor:115 - 145 µm
- Referencia:76500-654
- Membrana:Cellulose nitrate
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0.45 µm pore size are tested and released according to ISO 7704.