Orders LinkContactUsLinkComponent
Easy Sampler, Burkle
60 ml (2.03 oz.), 100 cm (39.37"), 32 mm (1.26")
  76501-528
 :   BURKLE INC
 :  5360-0600
Easy Sampler, Burkle
  76501-528
Easy Sampler, Burkle
Easy Sampler, Burkle
60 ml (2.03 oz.), 100 cm (39.37"), 32 mm (1.26")
$37,483.62
Each (60ml)
The Easy Sampler is designed to quickly and easily take multiple samples from bulk goods.
  • Stainless steel AISI 316 L (1.4404)
  • For sample openings from 52 mm (2.05") diameter
Emptying has been greatly simplified and in this model takes place via the tip. The sample can be emptied directly into the sample container without having to disassemble the device, in order to obtain a mixed sample for instance. The bulk material must be free-flowing or self-flowing.
  • Ø:
    32 mm (1.26")
  • Capacity:
    60 ml (2.03 oz.)
  • Length:
    100 cm (39.37")