SP Bel-Art Poxygrid® Bench-Top Biohazard Bag Holder Kit, Bel-Art Products, a Part of SP
Proveedor: Bel-Art Products, a Part of SP
Certificados
Acerca de este artículo
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
- Snug fitting polypropylene plastic cover helps hold the bag securely in place
- Reusable, can be cleaned by hand or in automated dish washers
- Autoclavable
The rectangular steel wire frame holds a polyethylene bag 22×28 cm (8¹/₂×11") and it is coated in easy to clean, durable, orange color epoxy.
Certificaciones: Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes.
Entrega: Kit includes 100 bench-top biohazard bags (11215-857); can also be used with Clavies autoclavable bags.
Frequently Bought Together