Orders LinkContactUsLinkComponent
VWR® Syringe Filters
VWR® Syringe Filters
VWR® Syringe Filters
Supplier:  VWR International
Certificates
VWR® Syringe Filters
8 Options Available Below
Product Details & Documents
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
  • Sterile blister packed filters are Ideal for sterile filtration
  • Nonsterile syringe filters are excellent for HPLC applications
  • Biosafe according to Class VI plastics tests
  • Available with Nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass fiber)
  • Available with hydrophilic or hydrophobic PTFE
  • Each filter has a female Luer-lock inlet and a male slip Luer outlet
Nylon and PTFE membranes are ideal for sample preparation and small volume chemical filtration.
Syringe filters with CA (cellulose acetate) or PES (polyether sulfone) membranes are ideal for sterile filtration of buffers, tissue culture media or additives, and other biologicals. The PES membrane offers high flow rates.
RC and PTFE hydrophilic type offer the best chemical resistance for both aqueous solutions and solvents.
Certifications: Manufactured under ISO9001:2015 quality management.
Caution: Hydrophobic PTFE is not intended for use with aqueous solutions.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 76479-034
  • 76479-004
  • 76479-008
  • 76479-048
  • 76479-052
  • 76479-040
  • 76479-044
  • 76478-980
  • Autoclavable:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
Especificaciones
Product Family Options
Nylon membrane
Información del producto
  • Ø
    Pore size
    Membrane
    Housing Material
    Sterility
    Availability
    Price
  • 25 mm
    0.22 µm
    Nylon
    PP
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $457.89
    $457.89
    Log in Para disponibilidad
    Case de 20 (100UN)
    Case de 20 (100UN) - $499.10
    $499.10
    Paquete de 100
       $457.89
    Case de 20 (100UN)
       $499.10
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-034
    Autoclavable
    Yes
PTFE membrane
Información del producto
  • Ø
    Pore size
    Membrane
    Housing Material
    Sterility
    Availability
    Price
  • 25 mm
    0.45 µm
    Hydrophobic PTFE
    PP
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $599.26
    $599.26
    Log in Para disponibilidad
    Case de 20 (100UN)
    Case de 20 (100UN) - $653.19
    $653.19
    Paquete de 100
       $599.26
    Case de 20 (100UN)
       $653.19
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-004
    Autoclavable
    Yes
  • 13 mm
    0.45 µm
    Hydrophobic PTFE
    PP
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $409.10
    $409.10
    Log in Para disponibilidad
    Case de 20 (100UN)
    Case de 20 (100UN) - $445.92
    $445.92
    Paquete de 100
       $409.10
    Case de 20 (100UN)
       $445.92
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-008
    Autoclavable
    Yes
  • 13 mm
    0.22 µm
    Hydrophilic PTFE
    PP
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $469.05
    $469.05
    Log in Para disponibilidad
    Case de 20 (100UN)
    Case de 20 (100UN) - $511.26
    $511.26
    Paquete de 100
       $469.05
    Case de 20 (100UN)
       $511.26
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-048
    Autoclavable
    Yes
  • 25 mm
    0.22 µm
    Hydrophilic PTFE
    PP
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $627.16
    $627.16
    Log in Para disponibilidad
    Case de 20 (100UN)
    Case de 20 (100UN) - $683.61
    $683.61
    Paquete de 100
       $627.16
    Case de 20 (100UN)
       $683.61
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-052
    Autoclavable
    Yes
CA membrane
Información del producto
  • Ø
    Pore size
    Membrane
    Housing Material
    Sterility
    Availability
    Price
  • 25 mm
    0.45 µm
    CA
    PP
    Sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 50
    Paquete de 50 - $488.43
    $488.43
    Log in Para disponibilidad
    Case de 10 (50UN)
    Case de 10 (50UN) - $532.39
    $532.39
    Paquete de 50
       $488.43
    Case de 10 (50UN)
       $532.39
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-040
    Autoclavable
    Yes
  • 25 mm
    0.22 µm
    CA
    PP
    Sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 50
    Paquete de 50 - $488.43
    $488.43
    Log in Para disponibilidad
    Case de 10 (50UN)
    Case de 10 (50UN) - $532.39
    $532.39
    Paquete de 50
       $488.43
    Case de 10 (50UN)
       $532.39
    Especificaciones:
    More
    Specifications
    Cat. No.
    76479-044
    Autoclavable
    Yes
RC membrane
Información del producto
  • Ø
    Pore size
    Membrane
    Housing Material
    Sterility
    Availability
    Price
  • 30 mm
    0.45 µm
    RC
    PP
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,631.75
    $1,631.75
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $1,778.61
    $1,778.61
    Paquete de 100
       $1,631.75
    Case de 10 (100UN)
       $1,778.61
    Especificaciones:
    More
    Specifications
    Cat. No.
    76478-980
    Autoclavable
    Yes