J.T.Baker® Plate Square Well
Supplier: Ritter, part of Avantor
J.T.Baker® Plate Square Well
Supplier: Ritter, part of Avantor
Certificates
Acerca de este artículo
These 96 deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
- Chemical resistant
- V-bottom well shape maximises liquid removal and aids resuspension
- No inner edges to interfere with magnetic bead collection
- Raised well rims to improve heat sealing
- Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilization.
3 Options Available Below
Product Details & Documents
These 96 deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
- Chemical resistant
- V-bottom well shape maximises liquid removal and aids resuspension
- No inner edges to interfere with magnetic bead collection
- Raised well rims to improve heat sealing
- Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilization.
Specifications for Product Family
Especificaciones
- Catalog No:
- 43001-0020
- 43001-0062
- 43001-0066
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Especificaciones
Product Family Options
Información del producto
- Well VolumeBottom StyleColorSterileAvailabilityPrice
- 43001-002096-well plateSupplier Number: 43001-00201Especificaciones:
Low Manufacturing Impact More
Specifications
Cat. No.43001-0020Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)Low Manufacturing Impact - 43001-006248-well plateSupplier Number: 43001-00621Especificaciones:
Low Manufacturing Impact More
Specifications
Cat. No.43001-0062Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)Low Manufacturing Impact - 43001-006624-well plateSupplier Number: 43001-00661Especificaciones:
Low Manufacturing Impact More
Specifications
Cat. No.43001-0066Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)Low Manufacturing Impact