Sterile Microcentrifuge and Centrifuge Tubes
Catalog # 470332-382
Supplier: MTC BIO, INC.
Specifications
- Esterilidad:Sterile
- Volumen:10 ml
- Descripción:Conical-bottom centrifuge tube, graduated
- Dimensiones:16×104 mm
- Referencia:470332-382
- Empaquetado:Bagged
- RCF máximo:10000×g
Specifications
About this item
These sterile tubes come in a variety of sizes and packaging options to fulfill a variety of tube needs in the lab.
- RNAse-, DNAse-, pyrogen-, and cytotoxin-free
- Store at −80 °C
- Sterilized by gamma radiation
These centrifuge tubes are perfect for everyday sample preparation needs. The rugged construction provides durable, reliable, long-term sample storage.