BioX Serological Pipettes
Serological pipette, 5 ml, Individually wrapped
Supplier: TRADEWINDS DIRECT INC
Número de proveedor #: BX07-5005
Jump to:
Product Details & Documents
These high quality serological pipettes are DNase/RNase free and come individually wrapped.
- Sterile
- Individually wrapped, paper/plastic wrap
- Non-pyrogenic
- DNase/RNase free
- Color coded
These sterile serological pipettes are not only DNase/RNase free and non pyrogenic, but they are also accurate to ±2% and color coded for your convenience.
Documents
Especificaciones
- Capacity:5 ml
- Description:Serological pipette
- Packed:Individually wrapped